Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39335811-39336080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMFG
Entrez Gene ID
9535
Gene Name
glia maturation factor gamma
Gene Aliases
GMF-GAMMA
UniGene
Hs.5210
Gene Chromosome Location
Chr.19: 39328357-39336086 on Build GRCh38
Transcripts
NM_001301008.1 NM_004877.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3701489 c.3+7G>A p.? Chr.19: 39335967-39335967 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs913177133 Chr.19: 39336040-39336040 on Build GRCh38
rs745952352 Chr.19: 39336020-39336020 on Build GRCh38
rs906504107 Chr.19: 39336029-39336029 on Build GRCh38
rs186000074 Chr.19: 39335902-39335902 on Build GRCh38
rs182038719 Chr.19: 39335883-39335883 on Build GRCh38
rs186330981 Chr.19: 39336051-39336051 on Build GRCh38
rs989595890 Chr.19: 39335838-39335838 on Build GRCh38
rs149387217 Chr.19: 39335975-39335975 on Build GRCh38
rs987579081 Chr.19: 39335874-39335874 on Build GRCh38
rs757445622 Chr.19: 39336010-39336010 on Build GRCh38
rs190607597 Chr.19: 39335861-39335861 on Build GRCh38
rs971328375 Chr.19: 39336045-39336045 on Build GRCh38
rs750420785 Chr.19: 39336054-39336054 on Build GRCh38
rs3786949 Chr.19: 39335839-39335839 on Build GRCh38
rs577538840 Chr.19: 39335870-39335870 on Build GRCh38
rs369627311 Chr.19: 39335918-39335918 on Build GRCh38
rs745902036 Chr.19: 39335933-39335933 on Build GRCh38
rs1038382240 Chr.19: 39336049-39336049 on Build GRCh38
rs368837888 Chr.19: 39335969-39335969 on Build GRCh38
rs369440333 Chr.19: 39336016-39336016 on Build GRCh38
rs568269395 Chr.19: 39336053-39336053 on Build GRCh38
rs757388319 Chr.19: 39336027-39336027 on Build GRCh38
rs574333378 Chr.19: 39335903-39335903 on Build GRCh38
rs560132092 Chr.19: 39336038-39336038 on Build GRCh38
rs1011532262 Chr.19: 39336052-39336052 on Build GRCh38
rs369601636 Chr.19: 39335939-39335939 on Build GRCh38
rs377300914 Chr.19: 39335991-39335991 on Build GRCh38
rs188954541 Chr.19: 39335966-39335966 on Build GRCh38
rs775463657 Chr.19: 39336028-39336028 on Build GRCh38
rs541483272 Chr.19: 39335914-39335914 on Build GRCh38
rs200197105 Chr.19: 39335960-39335960 on Build GRCh38
rs528296775 Chr.19: 39335954-39335954 on Build GRCh38
rs987201298 Chr.19: 39336056-39336056 on Build GRCh38
rs575950176 Chr.19: 39335984-39335984 on Build GRCh38
rs181638961 Chr.19: 39336048-39336048 on Build GRCh38
rs765908792 Chr.19: 39335965-39335965 on Build GRCh38
rs755616839 Chr.19: 39335961-39335961 on Build GRCh38
rs769740086 Chr.19: 39335934-39335934 on Build GRCh38
rs777972113 Chr.19: 39335992-39335992 on Build GRCh38
rs995898642 Chr.19: 39335976-39335976 on Build GRCh38
rs981874628 Chr.19: 39336046-39336047 on Build GRCh38
rs376637474 Chr.19: 39335935-39335935 on Build GRCh38
rs251905 Chr.19: 39335957-39335957 on Build GRCh38
rs372276903 Chr.19: 39335932-39335932 on Build GRCh38
rs563938601 Chr.19: 39336037-39336037 on Build GRCh38
rs759498683 Chr.19: 39335938-39335939 on Build GRCh38
rs966485153 Chr.19: 39335869-39335869 on Build GRCh38
rs763783990 Chr.19: 39335940-39335940 on Build GRCh38
rs372476128 Chr.19: 39335993-39335993 on Build GRCh38
rs34292370 Chr.19: 39335860-39335860 on Build GRCh38
rs376261266 Chr.19: 39336015-39336015 on Build GRCh38
rs769527080 Chr.19: 39336021-39336021 on Build GRCh38

View Full Product Details