Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6841453-6841726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNB3
Entrez Gene ID
2784
Gene Name
G protein subunit beta 3
Gene Aliases
CSNB1H
UniGene
Hs.631657
Gene Chromosome Location
Chr.12: 6840854-6847400 on Build GRCh38
Transcripts
NM_001297571.1 NM_002075.3 XM_011520953.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2099548 c.96+7C>T p.? Chr.12: 6841631-6841631 on Build GRCh38
1128481 c.59A>G p.D20G Chr.12: 6841587-6841587 on Build GRCh38
1165386 c.96+2T>A p.? Chr.12: 6841626-6841626 on Build GRCh38
695316 c.70G>C p.A24P Chr.12: 6841598-6841598 on Build GRCh38
5503867 c.60T>C p.D20D Chr.12: 6841588-6841588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782315249 Chr.12: 6841541-6841541 on Build GRCh38
rs982108818 Chr.12: 6841690-6841690 on Build GRCh38
rs782693345 Chr.12: 6841662-6841662 on Build GRCh38
rs782072645 Chr.12: 6841535-6841535 on Build GRCh38
rs13306407 Chr.12: 6841631-6841631 on Build GRCh38
rs782112871 Chr.12: 6841644-6841644 on Build GRCh38
rs938305715 Chr.12: 6841696-6841696 on Build GRCh38
rs781869004 Chr.12: 6841703-6841703 on Build GRCh38
rs913856195 Chr.12: 6841551-6841551 on Build GRCh38
rs782122055 Chr.12: 6841653-6841653 on Build GRCh38
rs782583806 Chr.12: 6841629-6841629 on Build GRCh38
rs782129332 Chr.12: 6841636-6841636 on Build GRCh38
rs145085793 Chr.12: 6841609-6841609 on Build GRCh38
rs781866961 Chr.12: 6841603-6841603 on Build GRCh38
rs782762128 Chr.12: 6841635-6841635 on Build GRCh38
rs782690581 Chr.12: 6841680-6841680 on Build GRCh38
rs782240155 Chr.12: 6841704-6841704 on Build GRCh38
rs561819792 Chr.12: 6841549-6841549 on Build GRCh38
rs782311919 Chr.12: 6841540-6841540 on Build GRCh38
rs781937681 Chr.12: 6841515-6841515 on Build GRCh38
rs1052437296 Chr.12: 6841518-6841518 on Build GRCh38
rs782499521 Chr.12: 6841664-6841664 on Build GRCh38
rs371123230 Chr.12: 6841640-6841640 on Build GRCh38
rs782463476 Chr.12: 6841626-6841626 on Build GRCh38
rs782784328 Chr.12: 6841602-6841602 on Build GRCh38
rs782605565 Chr.12: 6841504-6841504 on Build GRCh38
rs375208427 Chr.12: 6841666-6841666 on Build GRCh38
rs13306408 Chr.12: 6841577-6841577 on Build GRCh38
rs925338623 Chr.12: 6841496-6841496 on Build GRCh38
rs377388384 Chr.12: 6841679-6841679 on Build GRCh38
rs781801374 Chr.12: 6841659-6841659 on Build GRCh38
rs782102378 Chr.12: 6841559-6841559 on Build GRCh38
rs368336313 Chr.12: 6841657-6841657 on Build GRCh38
rs112239176 Chr.12: 6841705-6841705 on Build GRCh38
rs367711032 Chr.12: 6841567-6841567 on Build GRCh38
rs782820356 Chr.12: 6841547-6841547 on Build GRCh38
rs782014854 Chr.12: 6841650-6841650 on Build GRCh38
rs376340505 Chr.12: 6841655-6841655 on Build GRCh38
rs926859880 Chr.12: 6841694-6841694 on Build GRCh38
rs988593831 Chr.12: 6841672-6841672 on Build GRCh38
rs782534979 Chr.12: 6841701-6841701 on Build GRCh38
rs782294692 Chr.12: 6841632-6841632 on Build GRCh38
rs782562114 Chr.12: 6841600-6841600 on Build GRCh38
rs1002316792 Chr.12: 6841601-6841601 on Build GRCh38

View Full Product Details