Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67452156-67452415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP4
Entrez Gene ID
57010
Gene Name
calcium binding protein 4
Gene Aliases
CRSD, CSNB2B
UniGene
Hs.143036
Gene Chromosome Location
Chr.11: 67452406-67461774 on Build GRCh38
Transcripts
NM_001300896.1 XM_017018025.1 NM_206997.1

Gene Symbol
GPR152
Entrez Gene ID
390212
Gene Name
G protein-coupled receptor 152
Gene Aliases
PGR5
UniGene
Hs.567997
Gene Chromosome Location
Chr.11: 67451301-67452729 on Build GRCh38
Transcripts
NM_001300896.1 XM_017018025.1 NM_206997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3452305 c.532G>A p.D178N Chr.11: 67452193-67452193 on Build GRCh38
5899024 c.333G>A p.W111* Chr.11: 67452392-67452392 on Build GRCh38
1356428 c.351C>A p.S117S Chr.11: 67452374-67452374 on Build GRCh38
1356427 c.476C>T p.A159V Chr.11: 67452249-67452249 on Build GRCh38
3452306 c.474G>A p.L158L Chr.11: 67452251-67452251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191782000 Chr.11: 67452340-67452340 on Build GRCh38
rs117903569 Chr.11: 67452326-67452326 on Build GRCh38
rs775139507 Chr.11: 67452224-67452224 on Build GRCh38
rs748868428 Chr.11: 67452226-67452226 on Build GRCh38
rs527749058 Chr.11: 67452266-67452266 on Build GRCh38
rs777873355 Chr.11: 67452200-67452200 on Build GRCh38
rs755001152 Chr.11: 67452178-67452178 on Build GRCh38
rs756679563 Chr.11: 67452277-67452277 on Build GRCh38
rs994064657 Chr.11: 67452235-67452235 on Build GRCh38
rs554577764 Chr.11: 67452344-67452344 on Build GRCh38
rs761631025 Chr.11: 67452240-67452240 on Build GRCh38
rs144630243 Chr.11: 67452252-67452252 on Build GRCh38
rs774285725 Chr.11: 67452389-67452389 on Build GRCh38
rs750390771 Chr.11: 67452176-67452176 on Build GRCh38
rs556561917 Chr.11: 67452242-67452242 on Build GRCh38
rs149109930 Chr.11: 67452215-67452215 on Build GRCh38
rs372634169 Chr.11: 67452355-67452355 on Build GRCh38
rs763037504 Chr.11: 67452323-67452323 on Build GRCh38
rs769833209 Chr.11: 67452383-67452383 on Build GRCh38
rs758635098 Chr.11: 67452194-67452194 on Build GRCh38
rs780722780 Chr.11: 67452279-67452279 on Build GRCh38
rs778580128 Chr.11: 67452293-67452293 on Build GRCh38
rs778093692 Chr.11: 67452388-67452388 on Build GRCh38
rs776535111 Chr.11: 67452374-67452374 on Build GRCh38
rs961215424 Chr.11: 67452234-67452234 on Build GRCh38
rs371602409 Chr.11: 67452274-67452274 on Build GRCh38
rs747752250 Chr.11: 67452356-67452356 on Build GRCh38
rs796337570 Chr.11: 67452276-67452277 on Build GRCh38
rs768083274 Chr.11: 67452291-67452291 on Build GRCh38
rs757544766 Chr.11: 67452269-67452269 on Build GRCh38
rs1013487922 Chr.11: 67452393-67452393 on Build GRCh38
rs374063857 Chr.11: 67452231-67452231 on Build GRCh38
rs773039556 Chr.11: 67452245-67452245 on Build GRCh38
rs780669506 Chr.11: 67452343-67452343 on Build GRCh38
rs748564423 Chr.11: 67452267-67452267 on Build GRCh38
rs367720617 Chr.11: 67452327-67452327 on Build GRCh38
rs201188198 Chr.11: 67452197-67452197 on Build GRCh38
rs746764243 Chr.11: 67452370-67452370 on Build GRCh38
rs776578974 Chr.11: 67452318-67452318 on Build GRCh38
rs560127653 Chr.11: 67452265-67452265 on Build GRCh38
rs145363353 Chr.11: 67452339-67452339 on Build GRCh38
rs745757748 Chr.11: 67452379-67452379 on Build GRCh38
rs554436929 Chr.11: 67452264-67452264 on Build GRCh38
rs758553167 Chr.11: 67452336-67452336 on Build GRCh38
rs906502285 Chr.11: 67452190-67452190 on Build GRCh38
rs752936476 Chr.11: 67452310-67452312 on Build GRCh38
rs760600715 Chr.11: 67452248-67452248 on Build GRCh38
rs138530126 Chr.11: 67452288-67452288 on Build GRCh38
rs755343622 Chr.11: 67452276-67452277 on Build GRCh38
rs368998457 Chr.11: 67452205-67452205 on Build GRCh38
rs771653529 Chr.11: 67452364-67452364 on Build GRCh38
rs200792057 Chr.11: 67452298-67452298 on Build GRCh38
rs536577603 Chr.11: 67452206-67452206 on Build GRCh38
rs376409557 Chr.11: 67452268-67452268 on Build GRCh38
rs777405785 Chr.11: 67452368-67452368 on Build GRCh38
rs1003504971 Chr.11: 67452192-67452192 on Build GRCh38
rs761970403 Chr.11: 67452391-67452391 on Build GRCh38
rs760475740 Chr.11: 67452308-67452308 on Build GRCh38
rs529951302 Chr.11: 67452214-67452214 on Build GRCh38
rs201669048 Chr.11: 67452193-67452193 on Build GRCh38
rs957375863 Chr.11: 67452315-67452315 on Build GRCh38
rs1055556848 Chr.11: 67452385-67452385 on Build GRCh38
rs374518560 Chr.11: 67452295-67452295 on Build GRCh38
rs765270446 Chr.11: 67452186-67452186 on Build GRCh38
rs773881133 Chr.11: 67452236-67452236 on Build GRCh38
rs771778277 Chr.11: 67452297-67452297 on Build GRCh38
rs770786693 Chr.11: 67452314-67452314 on Build GRCh38
rs148000171 Chr.11: 67452373-67452373 on Build GRCh38

View Full Product Details