Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122402675-122402948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM162A
Entrez Gene ID
26355
Gene Name
family with sequence similarity 162 member A
Gene Aliases
C3orf28, E2IG5, HGTD-P
UniGene
Hs.584881
Gene Chromosome Location
Chr.3: 122384176-122410114 on Build GRCh38
Transcripts
NM_014367.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4399269 c.48G>C p.R16S Chr.3: 122402773-122402773 on Build GRCh38
1484505 c.102G>C p.L34F Chr.3: 122402827-122402827 on Build GRCh38
1752847 c.56A>T p.E19V Chr.3: 122402781-122402781 on Build GRCh38
7313440 c.157+1G>T p.? Chr.3: 122402883-122402883 on Build GRCh38
1238831 c.157C>T p.R53C Chr.3: 122402882-122402882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778487652 Chr.3: 122402780-122402780 on Build GRCh38
rs375211869 Chr.3: 122402883-122402883 on Build GRCh38
rs757936349 Chr.3: 122402717-122402720 on Build GRCh38
rs755484459 Chr.3: 122402902-122402902 on Build GRCh38
rs1009201381 Chr.3: 122402873-122402873 on Build GRCh38
rs370866042 Chr.3: 122402735-122402736 on Build GRCh38
rs201624968 Chr.3: 122402776-122402776 on Build GRCh38
rs745565586 Chr.3: 122402870-122402870 on Build GRCh38
rs756712535 Chr.3: 122402720-122402720 on Build GRCh38
rs531678380 Chr.3: 122402784-122402784 on Build GRCh38
rs527285562 Chr.3: 122402855-122402855 on Build GRCh38
rs3838601 Chr.3: 122402716-122402717 on Build GRCh38
rs745896455 Chr.3: 122402714-122402714 on Build GRCh38
rs1036051638 Chr.3: 122402762-122402762 on Build GRCh38
rs780685971 Chr.3: 122402731-122402731 on Build GRCh38
rs930471281 Chr.3: 122402817-122402817 on Build GRCh38
rs772806583 Chr.3: 122402812-122402812 on Build GRCh38
rs561749087 Chr.3: 122402710-122402710 on Build GRCh38
rs754381682 Chr.3: 122402887-122402887 on Build GRCh38
rs571304875 Chr.3: 122402717-122402724 on Build GRCh38
rs199578714 Chr.3: 122402882-122402882 on Build GRCh38
rs760126469 Chr.3: 122402830-122402830 on Build GRCh38
rs940387586 Chr.3: 122402709-122402709 on Build GRCh38
rs17850692 Chr.3: 122402874-122402874 on Build GRCh38
rs752371117 Chr.3: 122402865-122402865 on Build GRCh38
rs768511837 Chr.3: 122402805-122402805 on Build GRCh38
rs762127324 Chr.3: 122402869-122402869 on Build GRCh38
rs566993558 Chr.3: 122402724-122402725 on Build GRCh38
rs770086231 Chr.3: 122402749-122402749 on Build GRCh38
rs781338294 Chr.3: 122402712-122402712 on Build GRCh38
rs988258345 Chr.3: 122402702-122402702 on Build GRCh38
rs778571663 Chr.3: 122402789-122402789 on Build GRCh38
rs368489889 Chr.3: 122402915-122402915 on Build GRCh38
rs748416519 Chr.3: 122402914-122402914 on Build GRCh38
rs11555937 Chr.3: 122402813-122402813 on Build GRCh38
rs368095194 Chr.3: 122402878-122402878 on Build GRCh38
rs779339366 Chr.3: 122402909-122402909 on Build GRCh38
rs749708701 Chr.3: 122402773-122402773 on Build GRCh38
rs759386513 Chr.3: 122402850-122402850 on Build GRCh38
rs939737241 Chr.3: 122402916-122402916 on Build GRCh38
rs776548425 Chr.3: 122402840-122402840 on Build GRCh38
rs373633927 Chr.3: 122402783-122402783 on Build GRCh38
rs771553925 Chr.3: 122402794-122402794 on Build GRCh38
rs770901241 Chr.3: 122402839-122402839 on Build GRCh38

View Full Product Details