Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 54939534-54939763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83B
Entrez Gene ID
222584
Gene Name
family with sequence similarity 83 member B
Gene Aliases
C6orf143
UniGene
Hs.152423
Gene Chromosome Location
Chr.6: 54846323-54945099 on Build GRCh38
Transcripts
NM_001010872.2 XM_006715022.3 XM_011514394.2 XM_011514395.2 XM_017010478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1284868 c.757G>T p.A253S Chr.6: 54939728-54939728 on Build GRCh38
742488 c.746C>T p.S249L Chr.6: 54939717-54939717 on Build GRCh38
7096124 c.763C>G p.L255V Chr.6: 54939734-54939734 on Build GRCh38
3355561 c.784A>G p.T262A Chr.6: 54939755-54939755 on Build GRCh38
385968 c.757G>A p.A253T Chr.6: 54939728-54939728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755574008 Chr.6: 54939596-54939596 on Build GRCh38
rs757147227 Chr.6: 54939693-54939693 on Build GRCh38
rs538962124 Chr.6: 54939699-54939699 on Build GRCh38
rs769338727 Chr.6: 54939689-54939689 on Build GRCh38
rs757438563 Chr.6: 54939657-54939657 on Build GRCh38
rs745919013 Chr.6: 54939680-54939680 on Build GRCh38
rs1033548779 Chr.6: 54939616-54939616 on Build GRCh38
rs777108033 Chr.6: 54939692-54939692 on Build GRCh38
rs774066652 Chr.6: 54939711-54939711 on Build GRCh38
rs567409739 Chr.6: 54939643-54939643 on Build GRCh38
rs150867132 Chr.6: 54939681-54939681 on Build GRCh38
rs201889250 Chr.6: 54939684-54939684 on Build GRCh38
rs993567824 Chr.6: 54939586-54939586 on Build GRCh38
rs141544107 Chr.6: 54939644-54939644 on Build GRCh38
rs747518231 Chr.6: 54939573-54939573 on Build GRCh38
rs779257839 Chr.6: 54939667-54939667 on Build GRCh38
rs1002623010 Chr.6: 54939603-54939603 on Build GRCh38
rs889407217 Chr.6: 54939562-54939562 on Build GRCh38
rs193274102 Chr.6: 54939735-54939735 on Build GRCh38
rs762339351 Chr.6: 54939697-54939697 on Build GRCh38
rs930794598 Chr.6: 54939575-54939575 on Build GRCh38
rs115163660 Chr.6: 54939574-54939574 on Build GRCh38
rs572788200 Chr.6: 54939564-54939564 on Build GRCh38
rs550844631 Chr.6: 54939612-54939612 on Build GRCh38
rs767086906 Chr.6: 54939736-54939736 on Build GRCh38
rs747543916 Chr.6: 54939686-54939686 on Build GRCh38
rs1025500022 Chr.6: 54939594-54939594 on Build GRCh38
rs1009657623 Chr.6: 54939733-54939733 on Build GRCh38

View Full Product Details