Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130697485-130697744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC2
Entrez Gene ID
23404
Gene Name
exosome component 2
Gene Aliases
RRP4, Rrp4p, hRrp4p, p7
UniGene
Hs.654643
Gene Chromosome Location
Chr.9: 130693760-130704894 on Build GRCh38
Transcripts
NM_001282708.1 NM_001282709.1 NM_014285.6 NR_104230.1 XM_005272176.2 XM_006717023.2 XM_017014558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4682066 c.260G>A p.R87Q Chr.9: 130697617-130697617 on Build GRCh38
3329052 c.250G>A p.V84I Chr.9: 130697607-130697607 on Build GRCh38
6325257 c.249C>T p.I83I Chr.9: 130697606-130697606 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751628968 Chr.9: 130697641-130697641 on Build GRCh38
rs111747755 Chr.9: 130697552-130697552 on Build GRCh38
rs889296311 Chr.9: 130697568-130697568 on Build GRCh38
rs911641085 Chr.9: 130697590-130697590 on Build GRCh38
rs937341802 Chr.9: 130697685-130697685 on Build GRCh38
rs748372672 Chr.9: 130697616-130697616 on Build GRCh38
rs953029933 Chr.9: 130697535-130697535 on Build GRCh38
rs760612702 Chr.9: 130697619-130697619 on Build GRCh38
rs757774690 Chr.9: 130697554-130697554 on Build GRCh38
rs1006440166 Chr.9: 130697606-130697606 on Build GRCh38
rs146758009 Chr.9: 130697505-130697505 on Build GRCh38
rs367803499 Chr.9: 130697669-130697669 on Build GRCh38
rs778403601 Chr.9: 130697615-130697615 on Build GRCh38
rs1016475885 Chr.9: 130697610-130697610 on Build GRCh38
rs1049129299 Chr.9: 130697674-130697674 on Build GRCh38
rs770798940 Chr.9: 130697632-130697632 on Build GRCh38
rs758531351 Chr.9: 130697538-130697538 on Build GRCh38
rs752145475 Chr.9: 130697548-130697548 on Build GRCh38
rs368161060 Chr.9: 130697636-130697636 on Build GRCh38
rs779065288 Chr.9: 130697511-130697511 on Build GRCh38
rs756416784 Chr.9: 130697662-130697662 on Build GRCh38
rs776547436 Chr.9: 130697633-130697633 on Build GRCh38
rs182727844 Chr.9: 130697643-130697643 on Build GRCh38
rs1050593200 Chr.9: 130697564-130697564 on Build GRCh38
rs888415848 Chr.9: 130697680-130697680 on Build GRCh38
rs777952196 Chr.9: 130697543-130697543 on Build GRCh38
rs752883972 Chr.9: 130697640-130697640 on Build GRCh38
rs780421078 Chr.9: 130697663-130697663 on Build GRCh38
rs768417818 Chr.9: 130697599-130697599 on Build GRCh38
rs781702486 Chr.9: 130697566-130697566 on Build GRCh38
rs35380996 Chr.9: 130697716-130697716 on Build GRCh38
rs375990177 Chr.9: 130697661-130697661 on Build GRCh38
rs139286519 Chr.9: 130697617-130697617 on Build GRCh38
rs531053282 Chr.9: 130697635-130697635 on Build GRCh38
rs371298052 Chr.9: 130697645-130697645 on Build GRCh38

View Full Product Details