Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68290845-68291074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEM1B
Entrez Gene ID
10116
Gene Name
fem-1 homolog B
Gene Aliases
F1A-ALPHA, F1AA, FEM1-beta
UniGene
Hs.362733
Gene Chromosome Location
Chr.15: 68277803-68295865 on Build GRCh38
Transcripts
NM_015322.4 XM_017021859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3503449 c.1527T>C p.F509F Chr.15: 68290885-68290885 on Build GRCh38
5849639 c.1537C>T p.L513F Chr.15: 68290895-68290895 on Build GRCh38
964348 c.1707G>A p.T569T Chr.15: 68291065-68291065 on Build GRCh38
5849640 c.1644delT p.L550fs*1 Chr.15: 68291002-68291002 on Build GRCh38
1661834 c.1585G>C p.D529H Chr.15: 68290943-68290943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764464606 Chr.15: 68290947-68290947 on Build GRCh38
rs778055591 Chr.15: 68290927-68290927 on Build GRCh38
rs780106358 Chr.15: 68290976-68290976 on Build GRCh38
rs746907604 Chr.15: 68290986-68290986 on Build GRCh38
rs754065123 Chr.15: 68290939-68290939 on Build GRCh38
rs753925908 Chr.15: 68290961-68290961 on Build GRCh38
rs776079037 Chr.15: 68291045-68291045 on Build GRCh38
rs1006459250 Chr.15: 68291000-68291000 on Build GRCh38
rs901608331 Chr.15: 68290912-68290912 on Build GRCh38
rs761833966 Chr.15: 68290975-68290975 on Build GRCh38
rs776498279 Chr.15: 68290999-68290999 on Build GRCh38
rs181515066 Chr.15: 68290876-68290876 on Build GRCh38
rs772972196 Chr.15: 68291019-68291019 on Build GRCh38
rs769623041 Chr.15: 68291017-68291017 on Build GRCh38
rs762595379 Chr.15: 68291023-68291023 on Build GRCh38
rs745995263 Chr.15: 68290969-68290969 on Build GRCh38
rs747831755 Chr.15: 68291011-68291011 on Build GRCh38
rs200910706 Chr.15: 68290869-68290869 on Build GRCh38
rs142144377 Chr.15: 68290873-68290873 on Build GRCh38
rs1014865716 Chr.15: 68291013-68291013 on Build GRCh38
rs757432489 Chr.15: 68290940-68290940 on Build GRCh38
rs371624753 Chr.15: 68290918-68290918 on Build GRCh38
rs961911166 Chr.15: 68291038-68291038 on Build GRCh38
rs768506639 Chr.15: 68290987-68290987 on Build GRCh38
rs34021140 Chr.15: 68291044-68291044 on Build GRCh38

View Full Product Details