Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 3855960-3856233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLIS3
Entrez Gene ID
169792
Gene Name
GLIS family zinc finger 3
Gene Aliases
NDH, ZNF515
UniGene
Hs.162125
Gene Chromosome Location
Chr.9: 3824127-4310694 on Build GRCh38
Transcripts
NM_001042413.1 NM_152629.3 XM_005251386.4 XM_005251387.4 XM_005251388.4 XM_011517763.2 XM_011517764.2 XM_011517765.2 XM_011517766.2 XM_011517767.2 XM_017014361.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1109128 c.2306C>A p.P769H Chr.9: 3856176-3856176 on Build GRCh38
5977254 c.2471A>C p.H824P Chr.9: 3856011-3856011 on Build GRCh38
6487803 c.1974C>G p.N658K Chr.9: 3856043-3856043 on Build GRCh38
1109129 c.1841C>A p.P614H Chr.9: 3856176-3856176 on Build GRCh38
74763 c.1918C>G p.P640A Chr.9: 3856099-3856099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019213425 Chr.9: 3856209-3856209 on Build GRCh38
rs777777838 Chr.9: 3856069-3856069 on Build GRCh38
rs532996107 Chr.9: 3856078-3856078 on Build GRCh38
rs772106128 Chr.9: 3855994-3855994 on Build GRCh38
rs372221069 Chr.9: 3856062-3856062 on Build GRCh38
rs537966660 Chr.9: 3856144-3856144 on Build GRCh38
rs200694226 Chr.9: 3856143-3856143 on Build GRCh38
rs762226438 Chr.9: 3856028-3856028 on Build GRCh38
rs367576956 Chr.9: 3856151-3856151 on Build GRCh38
rs187615896 Chr.9: 3856122-3856122 on Build GRCh38
rs766607178 Chr.9: 3856031-3856031 on Build GRCh38
rs867985925 Chr.9: 3856108-3856108 on Build GRCh38
rs762977593 Chr.9: 3856010-3856010 on Build GRCh38
rs768706671 Chr.9: 3856177-3856177 on Build GRCh38
rs757230523 Chr.9: 3856155-3856155 on Build GRCh38
rs776849293 Chr.9: 3856090-3856090 on Build GRCh38
rs140981222 Chr.9: 3856011-3856011 on Build GRCh38
rs546482250 Chr.9: 3856086-3856086 on Build GRCh38
rs909967106 Chr.9: 3856048-3856048 on Build GRCh38
rs869320723 Chr.9: 3856143-3856144 on Build GRCh38
rs202074188 Chr.9: 3856045-3856045 on Build GRCh38
rs754727139 Chr.9: 3856051-3856051 on Build GRCh38
rs754312328 Chr.9: 3856132-3856132 on Build GRCh38
rs984193457 Chr.9: 3856014-3856014 on Build GRCh38
rs936028978 Chr.9: 3856102-3856102 on Build GRCh38
rs746756116 Chr.9: 3856076-3856076 on Build GRCh38
rs139958113 Chr.9: 3856169-3856169 on Build GRCh38
rs774232431 Chr.9: 3856117-3856117 on Build GRCh38
rs562001640 Chr.9: 3856006-3856006 on Build GRCh38
rs79816341 Chr.9: 3856114-3856114 on Build GRCh38
rs184373705 Chr.9: 3855997-3855997 on Build GRCh38
rs563920093 Chr.9: 3856061-3856061 on Build GRCh38
rs761305387 Chr.9: 3856129-3856129 on Build GRCh38
rs768078379 Chr.9: 3856030-3856030 on Build GRCh38
rs147473975 Chr.9: 3856142-3856142 on Build GRCh38
rs368773221 Chr.9: 3855996-3855996 on Build GRCh38
rs141417434 Chr.9: 3856059-3856059 on Build GRCh38
rs766830534 Chr.9: 3856131-3856131 on Build GRCh38
rs753512407 Chr.9: 3856047-3856047 on Build GRCh38
rs566474240 Chr.9: 3856118-3856118 on Build GRCh38
rs372118555 Chr.9: 3856190-3856190 on Build GRCh38
rs751465176 Chr.9: 3856152-3856152 on Build GRCh38
rs749485361 Chr.9: 3856095-3856095 on Build GRCh38
rs773477561 Chr.9: 3856191-3856191 on Build GRCh38
rs768977643 Chr.9: 3856107-3856107 on Build GRCh38
rs748140344 Chr.9: 3856184-3856184 on Build GRCh38
rs748013795 Chr.9: 3855990-3855990 on Build GRCh38
rs918596433 Chr.9: 3855999-3855999 on Build GRCh38
rs779220877 Chr.9: 3856181-3856181 on Build GRCh38
rs535058720 Chr.9: 3856126-3856126 on Build GRCh38
rs745671153 Chr.9: 3856159-3856159 on Build GRCh38
rs917384027 Chr.9: 3856075-3856075 on Build GRCh38
rs761845139 Chr.9: 3856154-3856154 on Build GRCh38
rs778527005 Chr.9: 3855988-3855988 on Build GRCh38
rs780948928 Chr.9: 3856157-3856157 on Build GRCh38
rs756573246 Chr.9: 3856034-3856034 on Build GRCh38

View Full Product Details