Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 46385851-46386124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRA2
Entrez Gene ID
2555
Gene Name
gamma-aminobutyric acid type A receptor alpha2 subunit
Gene Aliases
-
UniGene
Hs.116250
Gene Chromosome Location
Chr.4: 46243548-46390039 on Build GRCh38
Transcripts
NM_000807.2 NM_001114175.1 NM_001286827.1 XM_005248080.3 XM_011513675.1 XM_011513676.1 XM_017007982.1 XM_017007983.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7293722 c.62C>T p.T21M Chr.4: 46386102-46386102 on Build GRCh38
3392885 c.152A>C p.D51A Chr.4: 46386109-46386109 on Build GRCh38
4863290 c.70G>A p.G24S Chr.4: 46386094-46386094 on Build GRCh38
1055665 c.167G>A p.R56Q Chr.4: 46386094-46386094 on Build GRCh38
7293723 c.159C>T p.Y53Y Chr.4: 46386102-46386102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934580275 Chr.4: 46385899-46385899 on Build GRCh38
rs540349855 Chr.4: 46385922-46385922 on Build GRCh38
rs752693291 Chr.4: 46386090-46386090 on Build GRCh38
rs952210484 Chr.4: 46385904-46385904 on Build GRCh38
rs184899036 Chr.4: 46386042-46386042 on Build GRCh38
rs200089639 Chr.4: 46386043-46386043 on Build GRCh38
rs558679708 Chr.4: 46385995-46385995 on Build GRCh38
rs950170334 Chr.4: 46385963-46385963 on Build GRCh38
rs202138742 Chr.4: 46386047-46386047 on Build GRCh38
rs1023231853 Chr.4: 46385927-46385927 on Build GRCh38
rs756325973 Chr.4: 46386078-46386078 on Build GRCh38
rs752281984 Chr.4: 46385950-46385950 on Build GRCh38
rs1005526947 Chr.4: 46385974-46385974 on Build GRCh38
rs776046200 Chr.4: 46386028-46386028 on Build GRCh38
rs749596899 Chr.4: 46386083-46386083 on Build GRCh38
rs750482687 Chr.4: 46386059-46386059 on Build GRCh38
rs201219341 Chr.4: 46386046-46386046 on Build GRCh38
rs767677744 Chr.4: 46386048-46386048 on Build GRCh38
rs765251624 Chr.4: 46386088-46386088 on Build GRCh38
rs1044467430 Chr.4: 46385964-46385964 on Build GRCh38
rs201075932 Chr.4: 46386033-46386033 on Build GRCh38
rs200153605 Chr.4: 46386084-46386084 on Build GRCh38
rs897429811 Chr.4: 46386030-46386030 on Build GRCh38
rs11503015 Chr.4: 46386027-46386027 on Build GRCh38
rs905917369 Chr.4: 46385967-46385967 on Build GRCh38
rs370310540 Chr.4: 46386041-46386041 on Build GRCh38

View Full Product Details