Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165754420-165754689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT3
Entrez Gene ID
2591
Gene Name
polypeptide N-acetylgalactosaminyltransferase 3
Gene Aliases
GalNAc-T3, HFTC, HHS
UniGene
Hs.170986
Gene Chromosome Location
Chr.2: 165747803-165796352 on Build GRCh38
Transcripts
NM_004482.3 XM_005246449.1 XM_011510929.1 XM_017003770.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7326193 c.1595T>C p.M532T Chr.2: 165754658-165754658 on Build GRCh38
368765 c.1580G>T p.G527V Chr.2: 165754673-165754673 on Build GRCh38
3312710 c.1580G>A p.G527D Chr.2: 165754673-165754673 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760830864 Chr.2: 165754626-165754626 on Build GRCh38
rs41268661 Chr.2: 165754576-165754576 on Build GRCh38
rs199572087 Chr.2: 165754463-165754463 on Build GRCh38
rs371714077 Chr.2: 165754656-165754656 on Build GRCh38
rs764015322 Chr.2: 165754630-165754630 on Build GRCh38
rs766458456 Chr.2: 165754616-165754616 on Build GRCh38
rs907508066 Chr.2: 165754521-165754521 on Build GRCh38
rs1050928178 Chr.2: 165754520-165754520 on Build GRCh38
rs778435144 Chr.2: 165754619-165754619 on Build GRCh38
rs774391241 Chr.2: 165754621-165754621 on Build GRCh38
rs551939201 Chr.2: 165754544-165754544 on Build GRCh38
rs753948743 Chr.2: 165754644-165754644 on Build GRCh38
rs769480810 Chr.2: 165754583-165754583 on Build GRCh38
rs921371134 Chr.2: 165754481-165754481 on Build GRCh38
rs867896147 Chr.2: 165754534-165754534 on Build GRCh38
rs747685834 Chr.2: 165754582-165754582 on Build GRCh38
rs67722214 Chr.2: 165754463-165754463 on Build GRCh38
rs773197010 Chr.2: 165754597-165754597 on Build GRCh38
rs62174818 Chr.2: 165754496-165754496 on Build GRCh38
rs13010715 Chr.2: 165754546-165754546 on Build GRCh38
rs776356584 Chr.2: 165754578-165754578 on Build GRCh38
rs765424119 Chr.2: 165754647-165754647 on Build GRCh38
rs1050887178 Chr.2: 165754514-165754514 on Build GRCh38
rs200815957 Chr.2: 165754648-165754648 on Build GRCh38
rs762818320 Chr.2: 165754604-165754604 on Build GRCh38
rs890992915 Chr.2: 165754542-165754542 on Build GRCh38

View Full Product Details