Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75312717-75312968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FILIP1
Entrez Gene ID
27145
Gene Name
filamin A interacting protein 1
Gene Aliases
FILIP
UniGene
Hs.696158
Gene Chromosome Location
Chr.6: 75294895-75493829 on Build GRCh38
Transcripts
NM_001289987.1 NM_001300866.1 NM_015687.3 XM_005248713.3 XM_005248715.4 XM_011535756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
268127 c.3038C>G p.S1013C Chr.6: 75312794-75312794 on Build GRCh38
5522698 c.3081C>T p.S1027S Chr.6: 75312751-75312751 on Build GRCh38
3629901 c.2963G>A p.R988K Chr.6: 75312869-75312869 on Build GRCh38
3629902 c.2963G>A p.R988K Chr.6: 75312869-75312869 on Build GRCh38
6988902 c.3064G>A p.A1022T Chr.6: 75312768-75312768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754595660 Chr.6: 75312796-75312796 on Build GRCh38
rs200776430 Chr.6: 75312910-75312910 on Build GRCh38
rs765850763 Chr.6: 75312898-75312898 on Build GRCh38
rs757941734 Chr.6: 75312783-75312783 on Build GRCh38
rs749359869 Chr.6: 75312813-75312813 on Build GRCh38
rs200562337 Chr.6: 75312790-75312790 on Build GRCh38
rs753081715 Chr.6: 75312929-75312929 on Build GRCh38
rs781567238 Chr.6: 75312799-75312799 on Build GRCh38
rs778465988 Chr.6: 75312811-75312811 on Build GRCh38
rs756597851 Chr.6: 75312930-75312930 on Build GRCh38
rs528225863 Chr.6: 75312810-75312810 on Build GRCh38
rs762448746 Chr.6: 75312889-75312889 on Build GRCh38
rs750407136 Chr.6: 75312861-75312861 on Build GRCh38
rs780739578 Chr.6: 75312923-75312923 on Build GRCh38
rs370253937 Chr.6: 75312816-75312816 on Build GRCh38
rs749891762 Chr.6: 75312778-75312778 on Build GRCh38
rs767566641 Chr.6: 75312771-75312771 on Build GRCh38
rs757124319 Chr.6: 75312759-75312759 on Build GRCh38
rs962312690 Chr.6: 75312840-75312840 on Build GRCh38
rs765178508 Chr.6: 75312762-75312762 on Build GRCh38
rs148756313 Chr.6: 75312738-75312738 on Build GRCh38
rs776433993 Chr.6: 75312841-75312841 on Build GRCh38
rs34807169 Chr.6: 75312825-75312825 on Build GRCh38
rs201450409 Chr.6: 75312800-75312800 on Build GRCh38
rs567751339 Chr.6: 75312757-75312757 on Build GRCh38
rs372671237 Chr.6: 75312849-75312849 on Build GRCh38
rs868522742 Chr.6: 75312758-75312758 on Build GRCh38
rs746745541 Chr.6: 75312795-75312795 on Build GRCh38
rs751049238 Chr.6: 75312899-75312899 on Build GRCh38
rs774430800 Chr.6: 75312819-75312819 on Build GRCh38
rs377504805 Chr.6: 75312860-75312860 on Build GRCh38

View Full Product Details