Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 190860631-190860902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMNC
Entrez Gene ID
647309
Gene Name
geminin coiled-coil domain containing
Gene Aliases
GEMC1
UniGene
Hs.518490
Gene Chromosome Location
Chr.3: 190843271-190864200 on Build GRCh38
Transcripts
NM_001146686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3664618 c.104C>G p.T35R Chr.3: 190860758-190860758 on Build GRCh38
3333345 c.160C>A p.Q54K Chr.3: 190860702-190860702 on Build GRCh38
5531757 c.139C>T p.L47L Chr.3: 190860723-190860723 on Build GRCh38
3333347 c.62C>T p.P21L Chr.3: 190860800-190860800 on Build GRCh38
7369616 c.84A>T p.E28D Chr.3: 190860778-190860778 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs13075089 Chr.3: 190860777-190860777 on Build GRCh38
rs751308175 Chr.3: 190860738-190860738 on Build GRCh38
rs769490773 Chr.3: 190860831-190860831 on Build GRCh38
rs560192430 Chr.3: 190860757-190860757 on Build GRCh38
rs542619794 Chr.3: 190860759-190860759 on Build GRCh38
rs201469606 Chr.3: 190860800-190860800 on Build GRCh38
rs527883874 Chr.3: 190860804-190860804 on Build GRCh38
rs780369337 Chr.3: 190860874-190860875 on Build GRCh38
rs762195190 Chr.3: 190860878-190860878 on Build GRCh38
rs567758187 Chr.3: 190860876-190860876 on Build GRCh38
rs948246238 Chr.3: 190860725-190860725 on Build GRCh38
rs577398682 Chr.3: 190860720-190860720 on Build GRCh38
rs750015445 Chr.3: 190860685-190860685 on Build GRCh38
rs957064614 Chr.3: 190860873-190860873 on Build GRCh38
rs749453426 Chr.3: 190860765-190860765 on Build GRCh38
rs941332813 Chr.3: 190860784-190860784 on Build GRCh38
rs539982113 Chr.3: 190860722-190860722 on Build GRCh38
rs772537968 Chr.3: 190860875-190860875 on Build GRCh38
rs766318707 Chr.3: 190860734-190860734 on Build GRCh38
rs766265700 Chr.3: 190860880-190860880 on Build GRCh38
rs372354454 Chr.3: 190860799-190860799 on Build GRCh38
rs751854103 Chr.3: 190860877-190860877 on Build GRCh38
rs1031542561 Chr.3: 190860862-190860862 on Build GRCh38
rs778857469 Chr.3: 190860768-190860768 on Build GRCh38
rs757561341 Chr.3: 190860766-190860766 on Build GRCh38
rs761367896 Chr.3: 190860818-190860818 on Build GRCh38
rs183159491 Chr.3: 190860780-190860780 on Build GRCh38
rs954369018 Chr.3: 190860711-190860711 on Build GRCh38
rs376955349 Chr.3: 190860824-190860824 on Build GRCh38
rs998822632 Chr.3: 190860836-190860836 on Build GRCh38
rs1031301933 Chr.3: 190860658-190860658 on Build GRCh38
rs920461451 Chr.3: 190860776-190860776 on Build GRCh38
rs547963993 Chr.3: 190860805-190860805 on Build GRCh38
rs1032901606 Chr.3: 190860847-190860847 on Build GRCh38
rs999593648 Chr.3: 190860838-190860838 on Build GRCh38
rs190950211 Chr.3: 190860758-190860758 on Build GRCh38
rs368194675 Chr.3: 190860778-190860778 on Build GRCh38
rs1046420246 Chr.3: 190860815-190860815 on Build GRCh38
rs571392237 Chr.3: 190860721-190860721 on Build GRCh38
rs777110569 Chr.3: 190860811-190860811 on Build GRCh38
rs368708295 Chr.3: 190860681-190860681 on Build GRCh38
rs544071700 Chr.3: 190860675-190860675 on Build GRCh38

View Full Product Details