Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154532923-154533156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PD
Entrez Gene ID
2539
Gene Name
glucose-6-phosphate dehydrogenase
Gene Aliases
G6PD1
UniGene
Hs.461047
Gene Chromosome Location
Chr.X: 154531390-154547586 on Build GRCh38
Transcripts
NM_000402.4 NM_001042351.2

SNP Detail

SNP ID Chromosome Location
rs137852327 Chr.X: 154533122-154533122 on Build GRCh38
rs1021454151 Chr.X: 154533007-154533007 on Build GRCh38
rs76368830 Chr.X: 154532967-154532967 on Build GRCh38
rs398123544 Chr.X: 154532956-154532956 on Build GRCh38
rs75675507 Chr.X: 154533042-154533042 on Build GRCh38
rs76723693 Chr.X: 154533025-154533025 on Build GRCh38
rs782732731 Chr.X: 154533069-154533069 on Build GRCh38
rs782271022 Chr.X: 154533090-154533090 on Build GRCh38
rs782354645 Chr.X: 154532965-154532965 on Build GRCh38
rs369603624 Chr.X: 154532975-154532975 on Build GRCh38
rs782326802 Chr.X: 154533126-154533126 on Build GRCh38
rs199702562 Chr.X: 154533036-154533036 on Build GRCh38
rs782117333 Chr.X: 154532991-154532991 on Build GRCh38
rs781956763 Chr.X: 154532996-154532996 on Build GRCh38
rs587776730 Chr.X: 154533013-154533036 on Build GRCh38
rs782534709 Chr.X: 154533021-154533021 on Build GRCh38
rs5030869 Chr.X: 154532990-154532990 on Build GRCh38
rs781906610 Chr.X: 154533020-154533020 on Build GRCh38
rs34193178 Chr.X: 154532945-154532945 on Build GRCh38
rs201321423 Chr.X: 154532970-154532970 on Build GRCh38
rs782699946 Chr.X: 154533005-154533005 on Build GRCh38
rs370017540 Chr.X: 154533046-154533046 on Build GRCh38
rs150589041 Chr.X: 154533056-154533056 on Build GRCh38
rs781975796 Chr.X: 154533104-154533104 on Build GRCh38
rs782380777 Chr.X: 154533096-154533096 on Build GRCh38
rs782593777 Chr.X: 154533012-154533012 on Build GRCh38
rs782174983 Chr.X: 154532972-154532972 on Build GRCh38
rs963659791 Chr.X: 154533063-154533063 on Build GRCh38
rs75090888 Chr.X: 154532988-154532988 on Build GRCh38
rs782587403 Chr.X: 154533045-154533045 on Build GRCh38
rs782064465 Chr.X: 154533003-154533003 on Build GRCh38
rs137852347 Chr.X: 154533029-154533029 on Build GRCh38
rs782741566 Chr.X: 154532993-154532993 on Build GRCh38
rs137852339 Chr.X: 154533044-154533044 on Build GRCh38
rs137852342 Chr.X: 154532969-154532969 on Build GRCh38
rs781920344 Chr.X: 154533133-154533133 on Build GRCh38

View Full Product Details