Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 37002084-37002357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC8
Entrez Gene ID
11340
Gene Name
exosome component 8
Gene Aliases
CIP3, EAP2, OIP2, PCH1C, RRP43, Rrp43p, bA421P11.3, p9
UniGene
Hs.294041
Gene Chromosome Location
Chr.13: 37000373-37009614 on Build GRCh38
Transcripts
NM_181503.2 XM_006719763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6015310 c.54G>T p.L18L Chr.13: 37002309-37002309 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs77177263 Chr.13: 37002329-37002329 on Build GRCh38
rs551091674 Chr.13: 37002187-37002187 on Build GRCh38
rs188372980 Chr.13: 37002146-37002146 on Build GRCh38
rs941416679 Chr.13: 37002244-37002244 on Build GRCh38
rs745879466 Chr.13: 37002266-37002266 on Build GRCh38
rs769438130 Chr.13: 37002227-37002228 on Build GRCh38
rs745386104 Chr.13: 37002120-37002122 on Build GRCh38
rs762406784 Chr.13: 37002313-37002313 on Build GRCh38
rs774853638 Chr.13: 37002117-37002117 on Build GRCh38
rs768071810 Chr.13: 37002324-37002324 on Build GRCh38
rs535572723 Chr.13: 37002144-37002144 on Build GRCh38
rs865878647 Chr.13: 37002197-37002197 on Build GRCh38
rs762962778 Chr.13: 37002291-37002292 on Build GRCh38
rs900765216 Chr.13: 37002177-37002177 on Build GRCh38
rs919132434 Chr.13: 37002189-37002189 on Build GRCh38
rs748614024 Chr.13: 37002175-37002175 on Build GRCh38
rs566342827 Chr.13: 37002276-37002276 on Build GRCh38
rs942853893 Chr.13: 37002149-37002149 on Build GRCh38
rs201843398 Chr.13: 37002238-37002238 on Build GRCh38
rs773083535 Chr.13: 37002253-37002254 on Build GRCh38
rs771058109 Chr.13: 37002298-37002300 on Build GRCh38
rs765943212 Chr.13: 37002328-37002328 on Build GRCh38
rs1039333584 Chr.13: 37002128-37002128 on Build GRCh38
rs1006558333 Chr.13: 37002127-37002127 on Build GRCh38
rs368766707 Chr.13: 37002335-37002335 on Build GRCh38
rs774837831 Chr.13: 37002310-37002310 on Build GRCh38
rs748013483 Chr.13: 37002233-37002233 on Build GRCh38
rs377264694 Chr.13: 37002252-37002252 on Build GRCh38
rs755633867 Chr.13: 37002326-37002326 on Build GRCh38
rs776685222 Chr.13: 37002249-37002249 on Build GRCh38
rs750982862 Chr.13: 37002325-37002325 on Build GRCh38
rs533406536 Chr.13: 37002277-37002277 on Build GRCh38
rs764347456 Chr.13: 37002283-37002283 on Build GRCh38

View Full Product Details