Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77431234-77431507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNA14
Entrez Gene ID
9630
Gene Name
G protein subunit alpha 14
Gene Aliases
-
UniGene
Hs.657795
Gene Chromosome Location
Chr.9: 77423079-77648316 on Build GRCh38
Transcripts
NM_004297.3 XM_011519224.1 XM_017015341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3908202 c.543C>A p.P181P Chr.9: 77431371-77431371 on Build GRCh38
5411323 c.468C>T p.Y156Y Chr.9: 77431446-77431446 on Build GRCh38
6184076 c.542C>A p.P181H Chr.9: 77431372-77431372 on Build GRCh38
253724 c.489C>T p.I163I Chr.9: 77431425-77431425 on Build GRCh38
6672122 c.586A>G p.I196V Chr.9: 77431328-77431328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780471698 Chr.9: 77431340-77431340 on Build GRCh38
rs986009054 Chr.9: 77431304-77431304 on Build GRCh38
rs1041303258 Chr.9: 77431348-77431348 on Build GRCh38
rs771447300 Chr.9: 77431278-77431278 on Build GRCh38
rs773540430 Chr.9: 77431390-77431390 on Build GRCh38
rs998526944 Chr.9: 77431273-77431273 on Build GRCh38
rs754671726 Chr.9: 77431356-77431358 on Build GRCh38
rs958181017 Chr.9: 77431295-77431295 on Build GRCh38
rs563731081 Chr.9: 77431465-77431465 on Build GRCh38
rs545966792 Chr.9: 77431385-77431385 on Build GRCh38
rs199964099 Chr.9: 77431425-77431425 on Build GRCh38
rs765970014 Chr.9: 77431382-77431382 on Build GRCh38
rs762105673 Chr.9: 77431323-77431323 on Build GRCh38
rs368672064 Chr.9: 77431462-77431462 on Build GRCh38
rs772024460 Chr.9: 77431368-77431368 on Build GRCh38
rs140029377 Chr.9: 77431320-77431320 on Build GRCh38
rs776087157 Chr.9: 77431437-77431437 on Build GRCh38
rs760152396 Chr.9: 77431281-77431281 on Build GRCh38
rs763308392 Chr.9: 77431454-77431454 on Build GRCh38
rs375490549 Chr.9: 77431355-77431355 on Build GRCh38
rs775034382 Chr.9: 77431279-77431279 on Build GRCh38
rs373555513 Chr.9: 77431383-77431383 on Build GRCh38
rs189436617 Chr.9: 77431288-77431288 on Build GRCh38
rs754176053 Chr.9: 77431435-77431435 on Build GRCh38
rs543951531 Chr.9: 77431455-77431455 on Build GRCh38
rs201125691 Chr.9: 77431413-77431413 on Build GRCh38
rs746940639 Chr.9: 77431432-77431432 on Build GRCh38
rs772912335 Chr.9: 77431445-77431445 on Build GRCh38
rs369522795 Chr.9: 77431379-77431379 on Build GRCh38
rs147745034 Chr.9: 77431431-77431431 on Build GRCh38
rs761483742 Chr.9: 77431384-77431384 on Build GRCh38
rs925062604 Chr.9: 77431286-77431286 on Build GRCh38
rs902103114 Chr.9: 77431461-77431461 on Build GRCh38
rs752010673 Chr.9: 77431409-77431409 on Build GRCh38
rs28395813 Chr.9: 77431410-77431410 on Build GRCh38
rs770804772 Chr.9: 77431364-77431364 on Build GRCh38
rs758196305 Chr.9: 77431430-77431430 on Build GRCh38
rs752932358 Chr.9: 77431422-77431422 on Build GRCh38
rs142306459 Chr.9: 77431357-77431357 on Build GRCh38
rs765610407 Chr.9: 77431329-77431329 on Build GRCh38
rs758907366 Chr.9: 77431331-77431331 on Build GRCh38
rs61755086 Chr.9: 77431365-77431365 on Build GRCh38
rs552995324 Chr.9: 77431399-77431399 on Build GRCh38
rs759572884 Chr.9: 77431270-77431270 on Build GRCh38
rs761227446 Chr.9: 77431403-77431403 on Build GRCh38
rs139345882 Chr.9: 77431402-77431402 on Build GRCh38
rs781412050 Chr.9: 77431356-77431356 on Build GRCh38
rs371060420 Chr.9: 77431418-77431418 on Build GRCh38
rs769417405 Chr.9: 77431330-77431330 on Build GRCh38
rs769419048 Chr.9: 77431443-77431443 on Build GRCh38
rs778057978 Chr.9: 77431476-77431477 on Build GRCh38
rs28395811 Chr.9: 77431466-77431466 on Build GRCh38
rs757425340 Chr.9: 77431472-77431472 on Build GRCh38
rs201799120 Chr.9: 77431424-77431424 on Build GRCh38
rs748724376 Chr.9: 77431437-77431440 on Build GRCh38
rs775349997 Chr.9: 77431371-77431371 on Build GRCh38
rs753556211 Chr.9: 77431322-77431322 on Build GRCh38
rs745699154 Chr.9: 77431367-77431367 on Build GRCh38
rs774874925 Chr.9: 77431456-77431456 on Build GRCh38
rs751642002 Chr.9: 77431341-77431341 on Build GRCh38
rs745646033 Chr.9: 77431429-77431429 on Build GRCh38

View Full Product Details