Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3660014-3660287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFRA4
Entrez Gene ID
64096
Gene Name
GDNF family receptor alpha 4
Gene Aliases
-
UniGene
Hs.302025
Gene Chromosome Location
Chr.20: 3659292-3663399 on Build GRCh38
Transcripts
NM_022139.3 NM_145762.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
723805 c.802G>T p.E268* Chr.20: 3660175-3660175 on Build GRCh38
5724366 c.785A>G p.N262S Chr.20: 3660192-3660192 on Build GRCh38
6373778 c.763C>T p.P255S Chr.20: 3660214-3660214 on Build GRCh38
3405055 c.744C>A p.A248A Chr.20: 3660233-3660233 on Build GRCh38
2760739 c.793G>A p.G265R Chr.20: 3660184-3660184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919030306 Chr.20: 3660232-3660232 on Build GRCh38
rs375585615 Chr.20: 3660036-3660036 on Build GRCh38
rs563073540 Chr.20: 3660166-3660166 on Build GRCh38
rs545460535 Chr.20: 3660195-3660195 on Build GRCh38
rs755723552 Chr.20: 3660181-3660181 on Build GRCh38
rs752767048 Chr.20: 3660182-3660183 on Build GRCh38
rs111644613 Chr.20: 3660112-3660112 on Build GRCh38
rs769968310 Chr.20: 3660233-3660233 on Build GRCh38
rs558807968 Chr.20: 3660064-3660064 on Build GRCh38
rs553182220 Chr.20: 3660121-3660121 on Build GRCh38
rs1015997662 Chr.20: 3660257-3660257 on Build GRCh38
rs371337814 Chr.20: 3660156-3660156 on Build GRCh38
rs142300828 Chr.20: 3660218-3660218 on Build GRCh38
rs998373249 Chr.20: 3660106-3660106 on Build GRCh38
rs576933084 Chr.20: 3660096-3660096 on Build GRCh38
rs998061212 Chr.20: 3660208-3660208 on Build GRCh38
rs150162133 Chr.20: 3660220-3660220 on Build GRCh38
rs948115902 Chr.20: 3660077-3660077 on Build GRCh38
rs115995912 Chr.20: 3660098-3660098 on Build GRCh38
rs368122128 Chr.20: 3660252-3660252 on Build GRCh38
rs775564801 Chr.20: 3660234-3660234 on Build GRCh38
rs73893069 Chr.20: 3660126-3660126 on Build GRCh38
rs768332292 Chr.20: 3660189-3660189 on Build GRCh38
rs1028540707 Chr.20: 3660110-3660110 on Build GRCh38
rs374283828 Chr.20: 3660222-3660222 on Build GRCh38
rs762292479 Chr.20: 3660261-3660261 on Build GRCh38
rs1042506706 Chr.20: 3660081-3660081 on Build GRCh38
rs544481488 Chr.20: 3660185-3660185 on Build GRCh38
rs768100334 Chr.20: 3660266-3660266 on Build GRCh38
rs915763108 Chr.20: 3660074-3660074 on Build GRCh38
rs759582850 Chr.20: 3660226-3660226 on Build GRCh38
rs574392801 Chr.20: 3660123-3660123 on Build GRCh38
rs1054188235 Chr.20: 3660045-3660045 on Build GRCh38
rs1010047838 Chr.20: 3660073-3660073 on Build GRCh38
rs773161894 Chr.20: 3660224-3660224 on Build GRCh38
rs756510486 Chr.20: 3660108-3660109 on Build GRCh38
rs764226732 Chr.20: 3660243-3660243 on Build GRCh38
rs763306194 Chr.20: 3660240-3660240 on Build GRCh38
rs2853208 Chr.20: 3660176-3660176 on Build GRCh38
rs749026491 Chr.20: 3660187-3660187 on Build GRCh38

View Full Product Details