Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73434150-73434405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD9
Entrez Gene ID
8326
Gene Name
frizzled class receptor 9
Gene Aliases
CD349, FZD3
UniGene
Hs.647029
Gene Chromosome Location
Chr.7: 73433779-73436120 on Build GRCh38
Transcripts
NM_003508.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6362359 c.257G>A p.S86N Chr.7: 73434264-73434264 on Build GRCh38
5737954 c.326C>T p.P109L Chr.7: 73434333-73434333 on Build GRCh38
6668231 c.194C>T p.S65L Chr.7: 73434201-73434201 on Build GRCh38
5358303 c.365G>T p.C122F Chr.7: 73434372-73434372 on Build GRCh38
3762818 c.162G>C p.L54L Chr.7: 73434169-73434169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142125301 Chr.7: 73434334-73434334 on Build GRCh38
rs114148192 Chr.7: 73434214-73434214 on Build GRCh38
rs781814209 Chr.7: 73434327-73434327 on Build GRCh38
rs112773403 Chr.7: 73434357-73434357 on Build GRCh38
rs34455338 Chr.7: 73434352-73434353 on Build GRCh38
rs550209901 Chr.7: 73434289-73434289 on Build GRCh38
rs782377030 Chr.7: 73434379-73434379 on Build GRCh38
rs782000137 Chr.7: 73434383-73434383 on Build GRCh38
rs782199592 Chr.7: 73434294-73434294 on Build GRCh38
rs781911724 Chr.7: 73434250-73434250 on Build GRCh38
rs782164298 Chr.7: 73434199-73434199 on Build GRCh38
rs782389995 Chr.7: 73434178-73434178 on Build GRCh38
rs782704580 Chr.7: 73434326-73434326 on Build GRCh38
rs782695258 Chr.7: 73434206-73434206 on Build GRCh38
rs782739522 Chr.7: 73434238-73434238 on Build GRCh38
rs782600769 Chr.7: 73434374-73434374 on Build GRCh38
rs782689397 Chr.7: 73434167-73434167 on Build GRCh38
rs782523951 Chr.7: 73434343-73434343 on Build GRCh38
rs375005276 Chr.7: 73434355-73434355 on Build GRCh38
rs782757278 Chr.7: 73434333-73434333 on Build GRCh38
rs782343653 Chr.7: 73434295-73434295 on Build GRCh38
rs782249737 Chr.7: 73434366-73434367 on Build GRCh38
rs782015381 Chr.7: 73434191-73434191 on Build GRCh38
rs782476477 Chr.7: 73434329-73434329 on Build GRCh38
rs782174254 Chr.7: 73434352-73434352 on Build GRCh38
rs782111677 Chr.7: 73434311-73434311 on Build GRCh38
rs782213538 Chr.7: 73434271-73434271 on Build GRCh38
rs782328376 Chr.7: 73434172-73434172 on Build GRCh38
rs782103545 Chr.7: 73434177-73434177 on Build GRCh38
rs372060832 Chr.7: 73434252-73434252 on Build GRCh38
rs781854794 Chr.7: 73434239-73434239 on Build GRCh38
rs782410069 Chr.7: 73434166-73434166 on Build GRCh38
rs782797612 Chr.7: 73434315-73434315 on Build GRCh38
rs782165499 Chr.7: 73434322-73434322 on Build GRCh38
rs371558043 Chr.7: 73434344-73434344 on Build GRCh38
rs782268651 Chr.7: 73434286-73434286 on Build GRCh38
rs61749861 Chr.7: 73434169-73434169 on Build GRCh38
rs781964820 Chr.7: 73434174-73434174 on Build GRCh38
rs201878512 Chr.7: 73434255-73434255 on Build GRCh38
rs782016785 Chr.7: 73434320-73434320 on Build GRCh38
rs782634341 Chr.7: 73434272-73434272 on Build GRCh38
rs782231665 Chr.7: 73434376-73434376 on Build GRCh38
rs776325053 Chr.7: 73434345-73434345 on Build GRCh38
rs782079145 Chr.7: 73434237-73434237 on Build GRCh38
rs781972430 Chr.7: 73434301-73434301 on Build GRCh38
rs782798543 Chr.7: 73434245-73434245 on Build GRCh38

View Full Product Details