Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 25758061-25758286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABPA
Entrez Gene ID
2551
Gene Name
GA binding protein transcription factor alpha subunit
Gene Aliases
E4TF1-60, E4TF1A, NFT2, NRF2, NRF2A, RCH04A07
UniGene
Hs.473470
Gene Chromosome Location
Chr.21: 25734947-25772460 on Build GRCh38
Transcripts
NM_001197297.1 NM_002040.3 XM_005260938.4 XM_011529520.2 XM_011529521.2 XM_017028313.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1029623 c.653C>T p.S218L Chr.21: 25758109-25758109 on Build GRCh38
4572263 c.691T>A p.F231I Chr.21: 25758147-25758147 on Build GRCh38
1615934 c.673C>T p.L225F Chr.21: 25758129-25758129 on Build GRCh38
1307689 c.685G>T p.D229Y Chr.21: 25758141-25758141 on Build GRCh38
356957 c.685G>A p.D229N Chr.21: 25758141-25758141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147848277 Chr.21: 25758171-25758171 on Build GRCh38
rs971983400 Chr.21: 25758203-25758203 on Build GRCh38
rs759588641 Chr.21: 25758096-25758096 on Build GRCh38
rs753655053 Chr.21: 25758154-25758154 on Build GRCh38
rs112195834 Chr.21: 25758100-25758100 on Build GRCh38
rs1017993469 Chr.21: 25758233-25758233 on Build GRCh38
rs899587415 Chr.21: 25758248-25758248 on Build GRCh38
rs767605038 Chr.21: 25758098-25758098 on Build GRCh38
rs757936287 Chr.21: 25758163-25758163 on Build GRCh38
rs757068521 Chr.21: 25758155-25758155 on Build GRCh38
rs982943532 Chr.21: 25758213-25758213 on Build GRCh38
rs774456787 Chr.21: 25758215-25758216 on Build GRCh38
rs144668707 Chr.21: 25758162-25758162 on Build GRCh38
rs775561468 Chr.21: 25758245-25758245 on Build GRCh38
rs760955875 Chr.21: 25758110-25758110 on Build GRCh38
rs917399027 Chr.21: 25758179-25758179 on Build GRCh38
rs754641872 Chr.21: 25758207-25758207 on Build GRCh38
rs771332439 Chr.21: 25758228-25758228 on Build GRCh38
rs560721760 Chr.21: 25758218-25758218 on Build GRCh38
rs1052001003 Chr.21: 25758097-25758097 on Build GRCh38
rs550595053 Chr.21: 25758109-25758109 on Build GRCh38
rs774790322 Chr.21: 25758231-25758231 on Build GRCh38
rs765237436 Chr.21: 25758156-25758156 on Build GRCh38
rs746580753 Chr.21: 25758191-25758191 on Build GRCh38
rs763739223 Chr.21: 25758150-25758150 on Build GRCh38
rs979089135 Chr.21: 25758131-25758131 on Build GRCh38
rs760725634 Chr.21: 25758253-25758253 on Build GRCh38
rs375631254 Chr.21: 25758222-25758222 on Build GRCh38
rs368982441 Chr.21: 25758236-25758236 on Build GRCh38
rs1020897181 Chr.21: 25758103-25758103 on Build GRCh38
rs746404540 Chr.21: 25758234-25758234 on Build GRCh38

View Full Product Details