Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30741117-30741382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLOT1
Entrez Gene ID
10211
Gene Name
flotillin 1
Gene Aliases
-
UniGene
Hs.179986
Gene Chromosome Location
Chr.6: 30727709-30742851 on Build GRCh38
Transcripts
NM_001318875.1 NM_005803.3 XM_005248780.3 XM_006714947.3 XM_017010157.1 XM_017010158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
360162 c.269G>T p.G90V Chr.6: 30741275-30741275 on Build GRCh38
7414647 c.212T>C p.V71A Chr.6: 30741332-30741332 on Build GRCh38
3861535 c.308C>T p.T103M Chr.6: 30741236-30741236 on Build GRCh38
6665144 c.250G>A p.A84T Chr.6: 30741294-30741294 on Build GRCh38
281188 c.245C>A p.A82E Chr.6: 30741299-30741299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773964976 Chr.6: 30741279-30741279 on Build GRCh38
rs773805315 Chr.6: 30741137-30741137 on Build GRCh38
rs747028837 Chr.6: 30741249-30741249 on Build GRCh38
rs200920246 Chr.6: 30741244-30741244 on Build GRCh38
rs766012865 Chr.6: 30741313-30741313 on Build GRCh38
rs145255398 Chr.6: 30741295-30741295 on Build GRCh38
rs767034342 Chr.6: 30741142-30741142 on Build GRCh38
rs753497493 Chr.6: 30741360-30741360 on Build GRCh38
rs368750083 Chr.6: 30741186-30741186 on Build GRCh38
rs772669209 Chr.6: 30741243-30741243 on Build GRCh38
rs149998338 Chr.6: 30741269-30741269 on Build GRCh38
rs541162560 Chr.6: 30741298-30741298 on Build GRCh38
rs761767550 Chr.6: 30741294-30741294 on Build GRCh38
rs749079572 Chr.6: 30741267-30741267 on Build GRCh38
rs776911936 Chr.6: 30741265-30741265 on Build GRCh38
rs768162162 Chr.6: 30741250-30741250 on Build GRCh38
rs749891560 Chr.6: 30741144-30741144 on Build GRCh38
rs755764704 Chr.6: 30741163-30741163 on Build GRCh38
rs748713713 Chr.6: 30741202-30741202 on Build GRCh38
rs1059612 Chr.6: 30741178-30741178 on Build GRCh38
rs779326275 Chr.6: 30741181-30741181 on Build GRCh38
rs116542158 Chr.6: 30741167-30741167 on Build GRCh38
rs564230384 Chr.6: 30741318-30741318 on Build GRCh38
rs200270004 Chr.6: 30741211-30741211 on Build GRCh38

View Full Product Details