Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130763519-130763760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMER3
Entrez Gene ID
205147
Gene Name
APC membrane recruitment protein 3
Gene Aliases
FAM123C
UniGene
Hs.369289
Gene Chromosome Location
Chr.2: 130754475-130768134 on Build GRCh38
Transcripts
NM_001105193.1 NM_001105194.1 NM_001105195.1 NM_152698.2 XM_011510793.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5897364 c.1474G>A p.E492K Chr.2: 130763546-130763546 on Build GRCh38
6509754 c.1635G>A p.G545G Chr.2: 130763707-130763707 on Build GRCh38
5889696 c.1603G>A p.G535R Chr.2: 130763675-130763675 on Build GRCh38
5889695 c.1604G>A p.G535E Chr.2: 130763676-130763676 on Build GRCh38
6153947 c.1512C>A p.I504I Chr.2: 130763584-130763584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187906804 Chr.2: 130763740-130763740 on Build GRCh38
rs373842884 Chr.2: 130763610-130763610 on Build GRCh38
rs757763628 Chr.2: 130763709-130763709 on Build GRCh38
rs1050290292 Chr.2: 130763686-130763686 on Build GRCh38
rs749202289 Chr.2: 130763662-130763662 on Build GRCh38
rs902461607 Chr.2: 130763724-130763724 on Build GRCh38
rs1021319096 Chr.2: 130763567-130763567 on Build GRCh38
rs769412190 Chr.2: 130763657-130763657 on Build GRCh38
rs893863661 Chr.2: 130763703-130763704 on Build GRCh38
rs767583723 Chr.2: 130763604-130763604 on Build GRCh38
rs1047344373 Chr.2: 130763623-130763623 on Build GRCh38
rs773223965 Chr.2: 130763596-130763596 on Build GRCh38
rs754388093 Chr.2: 130763706-130763706 on Build GRCh38
rs370009156 Chr.2: 130763617-130763617 on Build GRCh38
rs747672659 Chr.2: 130763643-130763643 on Build GRCh38
rs377036438 Chr.2: 130763616-130763616 on Build GRCh38
rs376443283 Chr.2: 130763570-130763570 on Build GRCh38
rs746690951 Chr.2: 130763628-130763628 on Build GRCh38
rs1034859149 Chr.2: 130763729-130763729 on Build GRCh38
rs756385870 Chr.2: 130763734-130763735 on Build GRCh38
rs758994147 Chr.2: 130763578-130763578 on Build GRCh38
rs866640942 Chr.2: 130763699-130763699 on Build GRCh38
rs866095102 Chr.2: 130763717-130763717 on Build GRCh38
rs554983410 Chr.2: 130763688-130763688 on Build GRCh38
rs749660899 Chr.2: 130763562-130763562 on Build GRCh38
rs779306508 Chr.2: 130763627-130763627 on Build GRCh38
rs775633259 Chr.2: 130763694-130763694 on Build GRCh38
rs769857228 Chr.2: 130763590-130763590 on Build GRCh38
rs773346681 Chr.2: 130763658-130763658 on Build GRCh38
rs377413500 Chr.2: 130763741-130763741 on Build GRCh38
rs768458066 Chr.2: 130763579-130763579 on Build GRCh38
rs868593271 Chr.2: 130763712-130763712 on Build GRCh38
rs747613737 Chr.2: 130763587-130763587 on Build GRCh38
rs572440587 Chr.2: 130763613-130763613 on Build GRCh38
rs756634052 Chr.2: 130763545-130763545 on Build GRCh38
rs1003422928 Chr.2: 130763726-130763726 on Build GRCh38
rs554251959 Chr.2: 130763566-130763566 on Build GRCh38
rs200070399 Chr.2: 130763633-130763633 on Build GRCh38
rs764155520 Chr.2: 130763704-130763704 on Build GRCh38
rs778215262 Chr.2: 130763560-130763560 on Build GRCh38
rs759777563 Chr.2: 130763601-130763601 on Build GRCh38
rs753042159 Chr.2: 130763618-130763621 on Build GRCh38
rs750833774 Chr.2: 130763734-130763734 on Build GRCh38
rs373011055 Chr.2: 130763725-130763725 on Build GRCh38
rs369736958 Chr.2: 130763597-130763597 on Build GRCh38
rs752803111 Chr.2: 130763609-130763609 on Build GRCh38
rs770680817 Chr.2: 130763670-130763670 on Build GRCh38
rs758629744 Chr.2: 130763736-130763736 on Build GRCh38
rs776184160 Chr.2: 130763586-130763586 on Build GRCh38
rs765616041 Chr.2: 130763714-130763714 on Build GRCh38
rs774217643 Chr.2: 130763680-130763680 on Build GRCh38
rs941170710 Chr.2: 130763655-130763655 on Build GRCh38
rs767787582 Chr.2: 130763690-130763690 on Build GRCh38
rs779635155 Chr.2: 130763575-130763575 on Build GRCh38
rs757634081 Chr.2: 130763622-130763622 on Build GRCh38

View Full Product Details