Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 171868484-171868745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW11
Entrez Gene ID
23291
Gene Name
F-box and WD repeat domain containing 11
Gene Aliases
BTRC2, BTRCP2, FBW1B, FBXW1B, Fbw11, Hos
UniGene
Hs.484138
Gene Chromosome Location
Chr.5: 171861549-172006873 on Build GRCh38
Transcripts
NM_012300.2 NM_033644.2 NM_033645.2 XM_005265855.4 XM_005265856.2 XM_005265857.2 XM_005265858.2 XM_017009279.1 XM_017009280.1 XM_017009281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4768149 c.1622C>G p.S541C Chr.5: 171868642-171868642 on Build GRCh38
3614163 c.1597C>T p.P533S Chr.5: 171868667-171868667 on Build GRCh38
3853701 c.1520G>T p.S507I Chr.5: 171868744-171868744 on Build GRCh38
4395440 c.1532C>T p.P511L Chr.5: 171868693-171868693 on Build GRCh38
4684065 c.1552C>T p.R518C Chr.5: 171868673-171868673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376839553 Chr.5: 171868600-171868600 on Build GRCh38
rs956119138 Chr.5: 171868567-171868567 on Build GRCh38
rs901774781 Chr.5: 171868524-171868524 on Build GRCh38
rs376142107 Chr.5: 171868616-171868616 on Build GRCh38
rs192738546 Chr.5: 171868517-171868517 on Build GRCh38
rs61739670 Chr.5: 171868665-171868665 on Build GRCh38
rs745356602 Chr.5: 171868674-171868674 on Build GRCh38
rs746999494 Chr.5: 171868692-171868692 on Build GRCh38
rs368460351 Chr.5: 171868625-171868625 on Build GRCh38
rs571869925 Chr.5: 171868578-171868578 on Build GRCh38
rs997264568 Chr.5: 171868528-171868528 on Build GRCh38
rs778774761 Chr.5: 171868673-171868673 on Build GRCh38
rs759682790 Chr.5: 171868624-171868624 on Build GRCh38
rs547999783 Chr.5: 171868621-171868621 on Build GRCh38
rs773267748 Chr.5: 171868602-171868602 on Build GRCh38
rs775626358 Chr.5: 171868595-171868595 on Build GRCh38
rs116439645 Chr.5: 171868522-171868522 on Build GRCh38
rs766312829 Chr.5: 171868618-171868618 on Build GRCh38
rs144986711 Chr.5: 171868725-171868725 on Build GRCh38
rs150039691 Chr.5: 171868641-171868641 on Build GRCh38
rs761604414 Chr.5: 171868672-171868672 on Build GRCh38
rs756852927 Chr.5: 171868724-171868724 on Build GRCh38
rs910115938 Chr.5: 171868619-171868619 on Build GRCh38
rs756390635 Chr.5: 171868652-171868652 on Build GRCh38
rs746499270 Chr.5: 171868700-171868700 on Build GRCh38
rs763271487 Chr.5: 171868590-171868590 on Build GRCh38
rs764538744 Chr.5: 171868653-171868653 on Build GRCh38
rs371006606 Chr.5: 171868605-171868605 on Build GRCh38
rs1024442005 Chr.5: 171868549-171868549 on Build GRCh38
rs139563514 Chr.5: 171868688-171868688 on Build GRCh38
rs201693260 Chr.5: 171868597-171868597 on Build GRCh38
rs768287694 Chr.5: 171868722-171868722 on Build GRCh38

View Full Product Details