Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58569688-58569919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM107A
Entrez Gene ID
11170
Gene Name
family with sequence similarity 107 member A
Gene Aliases
DRR1, TU3A
UniGene
Hs.506357
Gene Chromosome Location
Chr.3: 58564112-58627610 on Build GRCh38
Transcripts
NM_001076778.2 NM_001282713.1 NM_001282714.1 NM_007177.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6544495 c.21G>A p.R7R Chr.3: 58569840-58569840 on Build GRCh38
4469446 c.15C>T p.I5I Chr.3: 58569846-58569846 on Build GRCh38
6658050 c.125G>A p.R42Q Chr.3: 58569736-58569736 on Build GRCh38
1651381 c.48G>T p.M16I Chr.3: 58569813-58569813 on Build GRCh38
5865177 c.107A>G p.N36S Chr.3: 58569754-58569754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770614363 Chr.3: 58569750-58569750 on Build GRCh38
rs140130365 Chr.3: 58569801-58569801 on Build GRCh38
rs764578729 Chr.3: 58569859-58569859 on Build GRCh38
rs796921828 Chr.3: 58569717-58569717 on Build GRCh38
rs765634778 Chr.3: 58569766-58569766 on Build GRCh38
rs771660928 Chr.3: 58569736-58569736 on Build GRCh38
rs770400859 Chr.3: 58569824-58569824 on Build GRCh38
rs749103121 Chr.3: 58569716-58569716 on Build GRCh38
rs748040936 Chr.3: 58569812-58569812 on Build GRCh38
rs910856092 Chr.3: 58569743-58569743 on Build GRCh38
rs375660727 Chr.3: 58569826-58569826 on Build GRCh38
rs745534613 Chr.3: 58569833-58569833 on Build GRCh38
rs774777238 Chr.3: 58569861-58569861 on Build GRCh38
rs748980396 Chr.3: 58569750-58569750 on Build GRCh38
rs763381481 Chr.3: 58569852-58569852 on Build GRCh38
rs765059452 Chr.3: 58569889-58569889 on Build GRCh38
rs148188719 Chr.3: 58569744-58569744 on Build GRCh38
rs764099823 Chr.3: 58569784-58569784 on Build GRCh38
rs921207953 Chr.3: 58569822-58569822 on Build GRCh38
rs752537978 Chr.3: 58569895-58569895 on Build GRCh38
rs376120787 Chr.3: 58569884-58569884 on Build GRCh38
rs758415230 Chr.3: 58569819-58569819 on Build GRCh38
rs748128493 Chr.3: 58569725-58569725 on Build GRCh38
rs751660770 Chr.3: 58569790-58569790 on Build GRCh38
rs11539084 Chr.3: 58569775-58569775 on Build GRCh38
rs758135519 Chr.3: 58569749-58569749 on Build GRCh38
rs757345690 Chr.3: 58569794-58569794 on Build GRCh38
rs772863622 Chr.3: 58569737-58569737 on Build GRCh38
rs974168323 Chr.3: 58569773-58569773 on Build GRCh38
rs746518448 Chr.3: 58569820-58569820 on Build GRCh38
rs11539085 Chr.3: 58569783-58569783 on Build GRCh38
rs545872613 Chr.3: 58569722-58569722 on Build GRCh38
rs746537453 Chr.3: 58569835-58569835 on Build GRCh38
rs775843558 Chr.3: 58569842-58569842 on Build GRCh38
rs750609554 Chr.3: 58569879-58569879 on Build GRCh38
rs11539087 Chr.3: 58569751-58569751 on Build GRCh38
rs919661376 Chr.3: 58569774-58569774 on Build GRCh38
rs779645856 Chr.3: 58569715-58569715 on Build GRCh38
rs11539091 Chr.3: 58569768-58569768 on Build GRCh38
rs11539090 Chr.3: 58569864-58569864 on Build GRCh38
rs373429682 Chr.3: 58569863-58569863 on Build GRCh38
rs550153059 Chr.3: 58569836-58569836 on Build GRCh38
rs960131861 Chr.3: 58569853-58569853 on Build GRCh38
rs778873071 Chr.3: 58569811-58569811 on Build GRCh38
rs754770704 Chr.3: 58569809-58569809 on Build GRCh38
rs200477209 Chr.3: 58569761-58569761 on Build GRCh38
rs11539089 Chr.3: 58569808-58569808 on Build GRCh38
rs112161868 Chr.3: 58569748-58569748 on Build GRCh38
rs768393615 Chr.3: 58569719-58569719 on Build GRCh38
rs903238971 Chr.3: 58569878-58569878 on Build GRCh38

View Full Product Details