Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 125307844-125308073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAPVD1
Entrez Gene ID
26130
Gene Name
GTPase activating protein and VPS9 domains 1
Gene Aliases
GAPEX5, GAPex-5, RAP6
UniGene
Hs.495134
Gene Chromosome Location
Chr.9: 125261794-125365011 on Build GRCh38
Transcripts
NM_001282679.1 NM_001282680.1 NM_001282681.1 NM_015635.3 XM_005251901.3 XM_005251904.3 XM_006717044.3 XM_011518499.2 XM_011518500.2 XM_011518502.2 XM_011518506.2 XM_011518507.2 XM_017014600.1 XM_017014601.1 XM_017014602.1 XM_017014603.1 XM_017014604.1 XM_017014605.1 XM_017014606.1 XM_017014607.1 XM_017014608.1 XM_017014609.1 XM_017014610.1 XM_017014611.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7100618 c.1412C>T p.P471L Chr.9: 125307851-125307851 on Build GRCh38
1105167 c.1430G>A p.R477Q Chr.9: 125307869-125307869 on Build GRCh38
1661946 c.1433T>C p.L478S Chr.9: 125307872-125307872 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768670134 Chr.9: 125307909-125307909 on Build GRCh38
rs780633434 Chr.9: 125307872-125307872 on Build GRCh38
rs141361170 Chr.9: 125307967-125307967 on Build GRCh38
rs774446745 Chr.9: 125307919-125307919 on Build GRCh38
rs773211915 Chr.9: 125307945-125307945 on Build GRCh38
rs761905937 Chr.9: 125307929-125307929 on Build GRCh38
rs949188555 Chr.9: 125307974-125307974 on Build GRCh38
rs549857793 Chr.9: 125307871-125307871 on Build GRCh38
rs759907408 Chr.9: 125307980-125307980 on Build GRCh38
rs543269450 Chr.9: 125308011-125308011 on Build GRCh38
rs745522909 Chr.9: 125307888-125307888 on Build GRCh38
rs1333055 Chr.9: 125307948-125307948 on Build GRCh38
rs766540171 Chr.9: 125307951-125307951 on Build GRCh38
rs772173839 Chr.9: 125307943-125307943 on Build GRCh38
rs769382954 Chr.9: 125307896-125307896 on Build GRCh38
rs199683264 Chr.9: 125307897-125307897 on Build GRCh38
rs80108869 Chr.9: 125307972-125307972 on Build GRCh38
rs58961746 Chr.9: 125307917-125307917 on Build GRCh38
rs555760517 Chr.9: 125307934-125307934 on Build GRCh38
rs942422674 Chr.9: 125308030-125308030 on Build GRCh38

View Full Product Details