Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52685998-52686243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNL3
Entrez Gene ID
26354
Gene Name
G protein nucleolar 3
Gene Aliases
C77032, E2IG3, NNP47, NS
UniGene
Hs.313544
Gene Chromosome Location
Chr.3: 52685920-52694497 on Build GRCh38
Transcripts
NM_014366.4 NM_206825.1 NM_206826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4687486 c.11C>A p.P4H Chr.3: 52686103-52686103 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs182211853 Chr.3: 52686200-52686200 on Build GRCh38
rs745722348 Chr.3: 52686075-52686075 on Build GRCh38
rs370536252 Chr.3: 52686115-52686115 on Build GRCh38
rs774330775 Chr.3: 52686106-52686106 on Build GRCh38
rs749217416 Chr.3: 52686096-52686096 on Build GRCh38
rs376018985 Chr.3: 52686068-52686068 on Build GRCh38
rs925710736 Chr.3: 52686031-52686031 on Build GRCh38
rs139726177 Chr.3: 52686142-52686142 on Build GRCh38
rs755470156 Chr.3: 52686066-52686066 on Build GRCh38
rs373856582 Chr.3: 52686072-52686072 on Build GRCh38
rs368025635 Chr.3: 52686103-52686103 on Build GRCh38
rs533292598 Chr.3: 52686038-52686038 on Build GRCh38
rs200394487 Chr.3: 52686058-52686058 on Build GRCh38
rs773579745 Chr.3: 52686117-52686117 on Build GRCh38
rs774955184 Chr.3: 52686083-52686084 on Build GRCh38
rs754182302 Chr.3: 52686138-52686138 on Build GRCh38
rs564137189 Chr.3: 52686037-52686037 on Build GRCh38
rs776460818 Chr.3: 52686045-52686045 on Build GRCh38
rs1006297854 Chr.3: 52686184-52686184 on Build GRCh38
rs943051191 Chr.3: 52686015-52686015 on Build GRCh38
rs945615428 Chr.3: 52686089-52686089 on Build GRCh38
rs1047557282 Chr.3: 52686177-52686177 on Build GRCh38
rs899270345 Chr.3: 52686169-52686169 on Build GRCh38
rs886518137 Chr.3: 52686020-52686020 on Build GRCh38
rs978291198 Chr.3: 52686049-52686049 on Build GRCh38
rs769789577 Chr.3: 52686055-52686055 on Build GRCh38
rs370709570 Chr.3: 52686185-52686185 on Build GRCh38
rs372215490 Chr.3: 52686149-52686149 on Build GRCh38
rs895125857 Chr.3: 52686207-52686207 on Build GRCh38
rs767813697 Chr.3: 52686059-52686059 on Build GRCh38
rs770567079 Chr.3: 52686043-52686043 on Build GRCh38
rs746504321 Chr.3: 52686070-52686070 on Build GRCh38
rs549660436 Chr.3: 52686077-52686077 on Build GRCh38
rs766879754 Chr.3: 52686125-52686125 on Build GRCh38
rs772514571 Chr.3: 52686064-52686064 on Build GRCh38
rs1108842 Chr.3: 52686064-52686064 on Build GRCh38
rs535617804 Chr.3: 52686065-52686065 on Build GRCh38
rs370184667 Chr.3: 52686071-52686071 on Build GRCh38
rs991451418 Chr.3: 52686039-52686039 on Build GRCh38
rs761944082 Chr.3: 52686111-52686111 on Build GRCh38
rs750879993 Chr.3: 52686060-52686060 on Build GRCh38
rs901769071 Chr.3: 52686170-52686170 on Build GRCh38
rs745634160 Chr.3: 52686048-52686048 on Build GRCh38
rs760912255 Chr.3: 52686124-52686124 on Build GRCh38
rs775394035 Chr.3: 52686056-52686056 on Build GRCh38
rs1016255755 Chr.3: 52686026-52686026 on Build GRCh38
rs779835399 Chr.3: 52686082-52686082 on Build GRCh38
rs374236086 Chr.3: 52686139-52686139 on Build GRCh38
rs1006351769 Chr.3: 52686024-52686024 on Build GRCh38
rs879406792 Chr.3: 52686100-52686100 on Build GRCh38
rs187868308 Chr.3: 52686083-52686083 on Build GRCh38
rs538527622 Chr.3: 52686171-52686171 on Build GRCh38
rs368535653 Chr.3: 52686143-52686143 on Build GRCh38
rs761138330 Chr.3: 52686208-52686208 on Build GRCh38
rs866838788 Chr.3: 52686181-52686181 on Build GRCh38
rs550657465 Chr.3: 52686033-52686033 on Build GRCh38

View Full Product Details