Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 127738933-127739198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXI2
Entrez Gene ID
399823
Gene Name
forkhead box I2
Gene Aliases
-
UniGene
Hs.449674
Gene Chromosome Location
Chr.10: 127737274-127741186 on Build GRCh38
Transcripts
NM_207426.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1146415 c.940G>T p.E314* Chr.10: 127738948-127738948 on Build GRCh38
7368642 c.938G>A p.R313Q Chr.10: 127738946-127738946 on Build GRCh38
5991883 c.386delG p.E130fs*>5 Chr.10: 127738946-127738946 on Build GRCh38
684051 c.388G>T p.E130* Chr.10: 127738948-127738948 on Build GRCh38
3435609 c.948C>T p.T316T Chr.10: 127738956-127738956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912198027 Chr.10: 127739072-127739072 on Build GRCh38
rs1033417843 Chr.10: 127739044-127739044 on Build GRCh38
rs757893269 Chr.10: 127738988-127738988 on Build GRCh38
rs373131983 Chr.10: 127738985-127738985 on Build GRCh38
rs750513664 Chr.10: 127739124-127739124 on Build GRCh38
rs1041918296 Chr.10: 127739107-127739107 on Build GRCh38
rs755679560 Chr.10: 127738996-127738996 on Build GRCh38
rs780826714 Chr.10: 127738991-127738991 on Build GRCh38
rs1011034813 Chr.10: 127739071-127739071 on Build GRCh38
rs910009010 Chr.10: 127739127-127739127 on Build GRCh38
rs906226936 Chr.10: 127739038-127739038 on Build GRCh38
rs976716385 Chr.10: 127739096-127739096 on Build GRCh38
rs533147899 Chr.10: 127739010-127739010 on Build GRCh38
rs941427522 Chr.10: 127739174-127739174 on Build GRCh38
rs748975491 Chr.10: 127739003-127739003 on Build GRCh38
rs1029569516 Chr.10: 127739098-127739098 on Build GRCh38
rs896489652 Chr.10: 127739126-127739126 on Build GRCh38
rs1020794938 Chr.10: 127739077-127739077 on Build GRCh38
rs985452363 Chr.10: 127739125-127739125 on Build GRCh38
rs376881710 Chr.10: 127738998-127738998 on Build GRCh38
rs201656253 Chr.10: 127738972-127738972 on Build GRCh38
rs200515796 Chr.10: 127738957-127738957 on Build GRCh38
rs966917400 Chr.10: 127739088-127739088 on Build GRCh38
rs749992805 Chr.10: 127738992-127738992 on Build GRCh38
rs144354907 Chr.10: 127739179-127739179 on Build GRCh38
rs551373207 Chr.10: 127739020-127739020 on Build GRCh38
rs764554847 Chr.10: 127738976-127738976 on Build GRCh38
rs759777246 Chr.10: 127738956-127738956 on Build GRCh38
rs752988740 Chr.10: 127738970-127738970 on Build GRCh38
rs762613364 Chr.10: 127738968-127738974 on Build GRCh38
rs1001806315 Chr.10: 127739041-127739041 on Build GRCh38
rs569770508 Chr.10: 127739116-127739116 on Build GRCh38
rs1039638917 Chr.10: 127739090-127739097 on Build GRCh38
rs768334353 Chr.10: 127739006-127739006 on Build GRCh38
rs893548383 Chr.10: 127739068-127739068 on Build GRCh38
rs1023827389 Chr.10: 127739180-127739180 on Build GRCh38
rs914506991 Chr.10: 127739091-127739091 on Build GRCh38
rs534328434 Chr.10: 127739120-127739121 on Build GRCh38

View Full Product Details