Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 74204947-74205182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPGT
Entrez Gene ID
8790
Gene Name
fucose-1-phosphate guanylyltransferase
Gene Aliases
GFPP
UniGene
Hs.480085
Gene Chromosome Location
Chr.1: 74198212-74208702 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1

Gene Symbol
FPGT-TNNI3K
Entrez Gene ID
100526835
Gene Name
FPGT-TNNI3K readthrough
Gene Aliases
-
UniGene
Hs.480085
Gene Chromosome Location
Chr.1: 74198212-74544432 on Build GRCh38
Transcripts
NM_001199328.2 NM_001199329.2 NM_003838.4 NM_001112808.2 NM_001199327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5891596 c.919G>T p.A307S Chr.1: 74204966-74204966 on Build GRCh38
6666634 c.933A>T p.G311G Chr.1: 74204980-74204980 on Build GRCh38
272699 c.1041_1043delTGT p.V350delV Chr.1: 74205088-74205090 on Build GRCh38
1296686 c.1095T>C p.Y365Y Chr.1: 74205142-74205142 on Build GRCh38
912069 c.969C>A p.V323V Chr.1: 74205016-74205016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968992185 Chr.1: 74205018-74205018 on Build GRCh38
rs1032601024 Chr.1: 74205052-74205052 on Build GRCh38
rs149465354 Chr.1: 74205042-74205042 on Build GRCh38
rs769505325 Chr.1: 74205066-74205066 on Build GRCh38
rs770607644 Chr.1: 74205043-74205043 on Build GRCh38
rs780234044 Chr.1: 74205020-74205020 on Build GRCh38
rs373692012 Chr.1: 74205089-74205089 on Build GRCh38
rs764593211 Chr.1: 74204992-74204992 on Build GRCh38
rs370662752 Chr.1: 74205029-74205029 on Build GRCh38
rs770812641 Chr.1: 74205081-74205081 on Build GRCh38
rs539785322 Chr.1: 74205009-74205009 on Build GRCh38
rs750584000 Chr.1: 74205011-74205011 on Build GRCh38
rs959261457 Chr.1: 74205057-74205057 on Build GRCh38
rs755096288 Chr.1: 74205153-74205153 on Build GRCh38
rs766713115 Chr.1: 74205133-74205133 on Build GRCh38
rs745470256 Chr.1: 74205059-74205059 on Build GRCh38
rs763397900 Chr.1: 74204969-74204969 on Build GRCh38
rs200131255 Chr.1: 74205056-74205056 on Build GRCh38
rs984116808 Chr.1: 74205114-74205114 on Build GRCh38
rs753913574 Chr.1: 74205034-74205034 on Build GRCh38
rs762425678 Chr.1: 74205095-74205095 on Build GRCh38
rs756394011 Chr.1: 74205017-74205017 on Build GRCh38
rs747779478 Chr.1: 74205021-74205023 on Build GRCh38
rs919135445 Chr.1: 74205147-74205147 on Build GRCh38
rs781081232 Chr.1: 74205053-74205053 on Build GRCh38
rs762452732 Chr.1: 74205067-74205067 on Build GRCh38
rs750825586 Chr.1: 74205128-74205128 on Build GRCh38
rs993374851 Chr.1: 74205063-74205063 on Build GRCh38
rs970100495 Chr.1: 74205083-74205083 on Build GRCh38
rs375346203 Chr.1: 74205152-74205152 on Build GRCh38
rs760752127 Chr.1: 74205132-74205132 on Build GRCh38
rs770489270 Chr.1: 74205088-74205090 on Build GRCh38
rs753980581 Chr.1: 74205024-74205024 on Build GRCh38
rs376392962 Chr.1: 74205134-74205134 on Build GRCh38
rs999113716 Chr.1: 74205023-74205023 on Build GRCh38
rs757552483 Chr.1: 74205000-74205000 on Build GRCh38
rs1032299727 Chr.1: 74205040-74205040 on Build GRCh38
rs767737331 Chr.1: 74205111-74205111 on Build GRCh38
rs950023049 Chr.1: 74205122-74205122 on Build GRCh38
rs981448250 Chr.1: 74205123-74205123 on Build GRCh38
rs779181619 Chr.1: 74205155-74205155 on Build GRCh38
rs768218682 Chr.1: 74205086-74205086 on Build GRCh38

View Full Product Details