Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69326877-69327114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFPT1
Entrez Gene ID
2673
Gene Name
glutamine--fructose-6-phosphate transaminase 1
Gene Aliases
CMS12, CMSTA1, GFA, GFAT, GFAT 1, GFAT1, GFAT1m, GFPT, GFPT1L, MSLG
UniGene
Hs.580300
Gene Chromosome Location
Chr.2: 69319769-69387254 on Build GRCh38
Transcripts
NM_001244710.1 NM_002056.3 XM_017003801.1 XM_017003802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6542683 c.1978C>A p.L660I Chr.2: 69326937-69326937 on Build GRCh38
4686806 c.1901C>G p.T634R Chr.2: 69327014-69327014 on Build GRCh38
3839863 c.1849G>T p.V617L Chr.2: 69327066-69327066 on Build GRCh38
3972319 c.1991G>A p.G664D Chr.2: 69326978-69326978 on Build GRCh38
6251080 c.1995C>A p.G665G Chr.2: 69326920-69326920 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758522540 Chr.2: 69327001-69327001 on Build GRCh38
rs781377676 Chr.2: 69327014-69327014 on Build GRCh38
rs368308347 Chr.2: 69327013-69327013 on Build GRCh38
rs772640415 Chr.2: 69327050-69327050 on Build GRCh38
rs868258818 Chr.2: 69326913-69326913 on Build GRCh38
rs759969980 Chr.2: 69326932-69326932 on Build GRCh38
rs371761341 Chr.2: 69327051-69327051 on Build GRCh38
rs778941720 Chr.2: 69327023-69327023 on Build GRCh38
rs770769786 Chr.2: 69327066-69327066 on Build GRCh38
rs776368106 Chr.2: 69327077-69327077 on Build GRCh38
rs768371999 Chr.2: 69327015-69327015 on Build GRCh38
rs538218189 Chr.2: 69326929-69326929 on Build GRCh38
rs766101867 Chr.2: 69326926-69326926 on Build GRCh38
rs775899275 Chr.2: 69327089-69327089 on Build GRCh38
rs751623973 Chr.2: 69327008-69327008 on Build GRCh38
rs202217663 Chr.2: 69327053-69327053 on Build GRCh38
rs753100765 Chr.2: 69326998-69326998 on Build GRCh38
rs772026265 Chr.2: 69327042-69327042 on Build GRCh38
rs765856240 Chr.2: 69326968-69326968 on Build GRCh38
rs777761159 Chr.2: 69327003-69327003 on Build GRCh38
rs17854372 Chr.2: 69326995-69326995 on Build GRCh38
rs368208403 Chr.2: 69327007-69327007 on Build GRCh38
rs760006760 Chr.2: 69327079-69327079 on Build GRCh38
rs765467067 Chr.2: 69327084-69327084 on Build GRCh38
rs937857366 Chr.2: 69326911-69326911 on Build GRCh38

View Full Product Details