Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 59264032-59264287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13C
Entrez Gene ID
220965
Gene Name
family with sequence similarity 13 member C
Gene Aliases
FAM13C1
UniGene
Hs.607594
Gene Chromosome Location
Chr.10: 59245933-59363186 on Build GRCh38
Transcripts
NM_001001971.2 NM_001143773.1 NM_001166698.1 NM_198215.3 XM_005269618.4 XM_005269619.4 XM_006717702.3 XM_006717703.3 XM_011539490.2 XM_011539491.2 XM_017015885.1 XM_017015886.1 XM_017015887.1 XM_017015888.1 XM_017015889.1 XM_017015890.1 XM_017015891.1 XM_017015892.1 XM_017015893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4814927 c.1010G>C p.R337P Chr.10: 59264099-59264099 on Build GRCh38
1474680 c.1010G>C p.R337P Chr.10: 59264099-59264099 on Build GRCh38
1702476 c.944C>T p.P315L Chr.10: 59264165-59264165 on Build GRCh38
3439135 c.987G>A p.W329* Chr.10: 59264122-59264122 on Build GRCh38
3807398 c.967A>C p.N323H Chr.10: 59264142-59264142 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753235458 Chr.10: 59264180-59264181 on Build GRCh38
rs371436185 Chr.10: 59264224-59264224 on Build GRCh38
rs772920908 Chr.10: 59264137-59264137 on Build GRCh38
rs990216153 Chr.10: 59264128-59264128 on Build GRCh38
rs137976326 Chr.10: 59264100-59264100 on Build GRCh38
rs544577803 Chr.10: 59264251-59264251 on Build GRCh38
rs71467058 Chr.10: 59264224-59264225 on Build GRCh38
rs538115420 Chr.10: 59264068-59264068 on Build GRCh38
rs748499961 Chr.10: 59264064-59264064 on Build GRCh38
rs375175726 Chr.10: 59264147-59264147 on Build GRCh38
rs780891400 Chr.10: 59264060-59264060 on Build GRCh38
rs766288018 Chr.10: 59264176-59264176 on Build GRCh38
rs771497792 Chr.10: 59264078-59264078 on Build GRCh38
rs778874102 Chr.10: 59264099-59264099 on Build GRCh38
rs755787039 Chr.10: 59264204-59264204 on Build GRCh38
rs376187928 Chr.10: 59264190-59264190 on Build GRCh38
rs762887683 Chr.10: 59264156-59264156 on Build GRCh38
rs538838463 Chr.10: 59264223-59264223 on Build GRCh38
rs752416394 Chr.10: 59264201-59264201 on Build GRCh38
rs368817959 Chr.10: 59264067-59264067 on Build GRCh38
rs556346617 Chr.10: 59264076-59264076 on Build GRCh38
rs71467059 Chr.10: 59264231-59264232 on Build GRCh38
rs372071096 Chr.10: 59264120-59264120 on Build GRCh38
rs749845319 Chr.10: 59264077-59264077 on Build GRCh38
rs759053855 Chr.10: 59264192-59264192 on Build GRCh38
rs767122140 Chr.10: 59264193-59264193 on Build GRCh38
rs35958081 Chr.10: 59264226-59264227 on Build GRCh38
rs769603713 Chr.10: 59264131-59264131 on Build GRCh38
rs751094395 Chr.10: 59264188-59264188 on Build GRCh38
rs766314585 Chr.10: 59264058-59264059 on Build GRCh38

View Full Product Details