Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 71283954-71284227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTR2
Entrez Gene ID
55783
Gene Name
cap methyltransferase 2
Gene Aliases
AFT, FTSJD1, HMTr2, MTr2
UniGene
Hs.72782
Gene Chromosome Location
Chr.16: 71282300-71290178 on Build GRCh38
Transcripts
NM_001099642.1 NM_001324374.1 NM_001324377.1 NM_001324378.1 NM_001324379.1 NM_018348.5 XM_011523228.1 XM_011523229.1 XM_017023444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6667664 c.1765C>T p.R589C Chr.16: 71284156-71284156 on Build GRCh38
5747757 c.1777_1778insA p.M593fs*14 Chr.16: 71284143-71284144 on Build GRCh38
6667663 c.1785A>G p.I595M Chr.16: 71284136-71284136 on Build GRCh38
3717053 c.1705G>T p.E569* Chr.16: 71284216-71284216 on Build GRCh38
3755033 c.1823C>A p.T608K Chr.16: 71284098-71284098 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768685586 Chr.16: 71284017-71284017 on Build GRCh38
rs745977271 Chr.16: 71284165-71284165 on Build GRCh38
rs748040644 Chr.16: 71284112-71284112 on Build GRCh38
rs778909409 Chr.16: 71284109-71284109 on Build GRCh38
rs758088029 Chr.16: 71284055-71284056 on Build GRCh38
rs772760499 Chr.16: 71284143-71284143 on Build GRCh38
rs200221398 Chr.16: 71284123-71284123 on Build GRCh38
rs3096381 Chr.16: 71284098-71284098 on Build GRCh38
rs767075469 Chr.16: 71284074-71284074 on Build GRCh38
rs377064558 Chr.16: 71284050-71284050 on Build GRCh38
rs747112942 Chr.16: 71284019-71284019 on Build GRCh38
rs777005890 Chr.16: 71284031-71284031 on Build GRCh38
rs754811555 Chr.16: 71284154-71284154 on Build GRCh38
rs774739666 Chr.16: 71284196-71284196 on Build GRCh38
rs545239850 Chr.16: 71284060-71284060 on Build GRCh38
rs749934259 Chr.16: 71284083-71284083 on Build GRCh38
rs749831737 Chr.16: 71284193-71284193 on Build GRCh38
rs763851216 Chr.16: 71284155-71284155 on Build GRCh38
rs373299938 Chr.16: 71284122-71284122 on Build GRCh38
rs756082420 Chr.16: 71284169-71284169 on Build GRCh38
rs772586675 Chr.16: 71284204-71284204 on Build GRCh38
rs149004042 Chr.16: 71284163-71284163 on Build GRCh38
rs749668675 Chr.16: 71284132-71284132 on Build GRCh38
rs146282486 Chr.16: 71284156-71284156 on Build GRCh38
rs142098464 Chr.16: 71284005-71284005 on Build GRCh38
rs940157417 Chr.16: 71284202-71284202 on Build GRCh38
rs766084517 Chr.16: 71284149-71284149 on Build GRCh38
rs189091912 Chr.16: 71284134-71284134 on Build GRCh38
rs543435476 Chr.16: 71284133-71284133 on Build GRCh38
rs368719534 Chr.16: 71284051-71284051 on Build GRCh38
rs755754653 Chr.16: 71284003-71284003 on Build GRCh38
rs748256920 Chr.16: 71284165-71284165 on Build GRCh38
rs771064200 Chr.16: 71284024-71284024 on Build GRCh38
rs780337305 Chr.16: 71284181-71284181 on Build GRCh38
rs376730118 Chr.16: 71283993-71283993 on Build GRCh38
rs768922763 Chr.16: 71284192-71284192 on Build GRCh38
rs749535167 Chr.16: 71284187-71284187 on Build GRCh38
rs759894524 Chr.16: 71284038-71284038 on Build GRCh38
rs201948334 Chr.16: 71284113-71284113 on Build GRCh38
rs769903092 Chr.16: 71284048-71284048 on Build GRCh38
rs946136809 Chr.16: 71283982-71283982 on Build GRCh38
rs769970271 Chr.16: 71284185-71284188 on Build GRCh38
rs749008810 Chr.16: 71284008-71284008 on Build GRCh38
rs748493382 Chr.16: 71284201-71284201 on Build GRCh38
rs760052126 Chr.16: 71284146-71284146 on Build GRCh38
rs868756180 Chr.16: 71284159-71284159 on Build GRCh38
rs781479698 Chr.16: 71284079-71284085 on Build GRCh38
rs141839766 Chr.16: 71284018-71284018 on Build GRCh38
rs755805975 Chr.16: 71284092-71284092 on Build GRCh38
rs764695090 Chr.16: 71284056-71284056 on Build GRCh38
rs781487255 Chr.16: 71284120-71284120 on Build GRCh38
rs762377510 Chr.16: 71284137-71284137 on Build GRCh38
rs753360412 Chr.16: 71284150-71284150 on Build GRCh38
rs750024387 Chr.16: 71283995-71283995 on Build GRCh38
rs902348543 Chr.16: 71284013-71284013 on Build GRCh38
rs754685718 Chr.16: 71284100-71284100 on Build GRCh38
rs774774000 Chr.16: 71284057-71284057 on Build GRCh38
rs777823715 Chr.16: 71284186-71284187 on Build GRCh38
rs760101348 Chr.16: 71284126-71284126 on Build GRCh38
rs756717787 Chr.16: 71283991-71283991 on Build GRCh38

View Full Product Details