Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 74451949-74452212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLG1
Entrez Gene ID
2734
Gene Name
golgi glycoprotein 1
Gene Aliases
CFR-1, ESL-1, MG-160, MG160
UniGene
Hs.109731
Gene Chromosome Location
Chr.16: 74447427-74607144 on Build GRCh38
Transcripts
NM_001145666.1 NM_001145667.1 NM_012201.5 NR_027264.1 NR_027265.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3957819 c.3537G>T p.R1179S Chr.16: 74452131-74452131 on Build GRCh38
6671454 c.3595A>G p.T1199A Chr.16: 74452073-74452073 on Build GRCh38
973621 c.3564T>C p.A1188A Chr.16: 74452104-74452104 on Build GRCh38
74755 c.3570A>C p.K1190N Chr.16: 74452098-74452098 on Build GRCh38
5385374 c.3588G>A p.Q1196Q Chr.16: 74452080-74452080 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1005627479 Chr.16: 74452068-74452068 on Build GRCh38
rs760202878 Chr.16: 74452132-74452132 on Build GRCh38
rs759527869 Chr.16: 74452153-74452153 on Build GRCh38
rs554648057 Chr.16: 74452021-74452021 on Build GRCh38
rs758217492 Chr.16: 74452174-74452174 on Build GRCh38
rs532723662 Chr.16: 74452117-74452117 on Build GRCh38
rs754782479 Chr.16: 74452071-74452071 on Build GRCh38
rs776324035 Chr.16: 74452151-74452151 on Build GRCh38
rs372420402 Chr.16: 74452091-74452091 on Build GRCh38
rs138842904 Chr.16: 74452127-74452127 on Build GRCh38
rs774013468 Chr.16: 74452044-74452044 on Build GRCh38
rs763247124 Chr.16: 74452038-74452038 on Build GRCh38
rs765281655 Chr.16: 74452162-74452162 on Build GRCh38
rs775982562 Chr.16: 74452034-74452034 on Build GRCh38
rs558415697 Chr.16: 74451985-74451985 on Build GRCh38
rs373377309 Chr.16: 74452026-74452026 on Build GRCh38
rs769028626 Chr.16: 74452041-74452041 on Build GRCh38
rs778823590 Chr.16: 74452084-74452084 on Build GRCh38
rs771227537 Chr.16: 74452016-74452016 on Build GRCh38
rs781544832 Chr.16: 74452098-74452098 on Build GRCh38
rs770281947 Chr.16: 74452031-74452031 on Build GRCh38
rs768010465 Chr.16: 74452167-74452167 on Build GRCh38
rs752590164 Chr.16: 74452089-74452089 on Build GRCh38
rs774556582 Chr.16: 74452122-74452122 on Build GRCh38
rs369902011 Chr.16: 74452007-74452007 on Build GRCh38
rs772918405 Chr.16: 74452130-74452130 on Build GRCh38
rs1028886982 Chr.16: 74452175-74452175 on Build GRCh38
rs760037421 Chr.16: 74452155-74452156 on Build GRCh38
rs774996382 Chr.16: 74452109-74452109 on Build GRCh38
rs372443954 Chr.16: 74452002-74452002 on Build GRCh38
rs558085495 Chr.16: 74452059-74452059 on Build GRCh38
rs150213948 Chr.16: 74452111-74452111 on Build GRCh38
rs544174334 Chr.16: 74452006-74452006 on Build GRCh38
rs766020272 Chr.16: 74452147-74452147 on Build GRCh38
rs546533936 Chr.16: 74452119-74452119 on Build GRCh38
rs574799698 Chr.16: 74452048-74452048 on Build GRCh38
rs1050645 Chr.16: 74452050-74452050 on Build GRCh38
rs1045273841 Chr.16: 74451987-74451987 on Build GRCh38
rs745869989 Chr.16: 74452101-74452101 on Build GRCh38
rs140976108 Chr.16: 74452067-74452067 on Build GRCh38
rs575254187 Chr.16: 74451994-74451994 on Build GRCh38
rs374970900 Chr.16: 74452169-74452169 on Build GRCh38
rs578176804 Chr.16: 74452066-74452066 on Build GRCh38
rs749883852 Chr.16: 74452057-74452057 on Build GRCh38
rs747228950 Chr.16: 74452009-74452009 on Build GRCh38
rs370303587 Chr.16: 74452128-74452128 on Build GRCh38
rs376037327 Chr.16: 74452003-74452003 on Build GRCh38
rs769190110 Chr.16: 74452120-74452120 on Build GRCh38
rs377537936 Chr.16: 74452047-74452047 on Build GRCh38

View Full Product Details