Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132862175-132862444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF9
Entrez Gene ID
2661
Gene Name
growth differentiation factor 9
Gene Aliases
-
UniGene
Hs.25022
Gene Chromosome Location
Chr.5: 132861181-132866884 on Build GRCh38
Transcripts
NM_001288824.2 NM_001288825.2 NM_001288826.2 NM_001288827.2 NM_001288828.2 NM_005260.4 XM_005271957.4 XM_006714585.3 XM_011543308.2 XM_011543309.1 XM_011543310.1 XM_011543311.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
448578 c.750G>C p.Q250H Chr.5: 132862204-132862204 on Build GRCh38
1060774 c.686G>T p.R229I Chr.5: 132862268-132862268 on Build GRCh38
3137649 c.689G>C p.S230T Chr.5: 132862265-132862265 on Build GRCh38
3137652 c.604C>T p.Q202* Chr.5: 132862350-132862350 on Build GRCh38
1433088 c.614T>A p.F205Y Chr.5: 132862340-132862340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021022494 Chr.5: 132862265-132862265 on Build GRCh38
rs747727006 Chr.5: 132862220-132862220 on Build GRCh38
rs749717118 Chr.5: 132862372-132862372 on Build GRCh38
rs770359022 Chr.5: 132862248-132862248 on Build GRCh38
rs776127098 Chr.5: 132862251-132862251 on Build GRCh38
rs754720328 Chr.5: 132862214-132862214 on Build GRCh38
rs761889208 Chr.5: 132862268-132862268 on Build GRCh38
rs746532187 Chr.5: 132862241-132862241 on Build GRCh38
rs972496912 Chr.5: 132862237-132862237 on Build GRCh38
rs774519449 Chr.5: 132862381-132862381 on Build GRCh38
rs757930738 Chr.5: 132862344-132862344 on Build GRCh38
rs753755902 Chr.5: 132862300-132862300 on Build GRCh38
rs139629952 Chr.5: 132862350-132862350 on Build GRCh38
rs927239486 Chr.5: 132862287-132862287 on Build GRCh38
rs972839725 Chr.5: 132862396-132862396 on Build GRCh38
rs769225393 Chr.5: 132862377-132862377 on Build GRCh38
rs777370243 Chr.5: 132862240-132862240 on Build GRCh38
rs768506285 Chr.5: 132862387-132862387 on Build GRCh38
rs770542892 Chr.5: 132862366-132862366 on Build GRCh38
rs1029833498 Chr.5: 132862412-132862412 on Build GRCh38
rs372887898 Chr.5: 132862201-132862201 on Build GRCh38
rs374872541 Chr.5: 132862276-132862276 on Build GRCh38
rs779660194 Chr.5: 132862271-132862271 on Build GRCh38
rs748546274 Chr.5: 132862383-132862383 on Build GRCh38
rs779967455 Chr.5: 132862202-132862202 on Build GRCh38
rs769779936 Chr.5: 132862219-132862219 on Build GRCh38
rs752561305 Chr.5: 132862324-132862324 on Build GRCh38
rs750431662 Chr.5: 132862274-132862274 on Build GRCh38
rs771789570 Chr.5: 132862236-132862236 on Build GRCh38
rs749069992 Chr.5: 132862297-132862297 on Build GRCh38
rs766507252 Chr.5: 132862296-132862296 on Build GRCh38
rs1010013865 Chr.5: 132862264-132862264 on Build GRCh38
rs774807342 Chr.5: 132862260-132862260 on Build GRCh38
rs760940033 Chr.5: 132862392-132862392 on Build GRCh38
rs10491279 Chr.5: 132862408-132862408 on Build GRCh38
rs753843694 Chr.5: 132862211-132862211 on Build GRCh38
rs777572094 Chr.5: 132862349-132862349 on Build GRCh38
rs144257913 Chr.5: 132862384-132862384 on Build GRCh38
rs150812210 Chr.5: 132862257-132862257 on Build GRCh38
rs754986069 Chr.5: 132862317-132862317 on Build GRCh38
rs1055501297 Chr.5: 132862328-132862328 on Build GRCh38
rs778534564 Chr.5: 132862323-132862323 on Build GRCh38
rs767789846 Chr.5: 132862269-132862269 on Build GRCh38

View Full Product Details