Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 27186390-27186647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD5
Entrez Gene ID
91452
Gene Name
acyl-CoA binding domain containing 5
Gene Aliases
-
UniGene
Hs.530597
Gene Chromosome Location
Chr.10: 27182624-27248674 on Build GRCh38
Transcripts
XM_017016884.1 XM_017016885.1 XM_017016887.1 XM_017016888.1 XM_017016889.1 XM_017016890.1 XM_017016891.1 XM_017016893.1 XM_017016895.1 XM_017016896.1 XM_017016898.1 XM_017016900.1 XM_017016901.1 XM_017016904.1 XM_017016905.1 NM_001172303.2 NM_001172304.2 NM_001320756.1 NM_001320757.1 NM_032844.4 NR_135469.1 XM_005252631.3 XM_005252632.3 XM_006717519.3 XM_011519751.2 XM_017016852.1 XM_017016853.1 XM_017016856.1

Gene Symbol
MASTL
Entrez Gene ID
84930
Gene Name
microtubule associated serine/threonine kinase like
Gene Aliases
GREATWALL, GW, GWL, MAST-L, THC2
UniGene
Hs.276905
Gene Chromosome Location
Chr.10: 27154468-27187953 on Build GRCh38
Transcripts
XM_017016884.1 XM_017016885.1 XM_017016887.1 XM_017016888.1 XM_017016889.1 XM_017016890.1 XM_017016891.1 XM_017016893.1 XM_017016895.1 XM_017016896.1 XM_017016898.1 XM_017016900.1 XM_017016901.1 XM_017016904.1 XM_017016905.1 NM_001172303.2 NM_001172304.2 NM_001320756.1 NM_001320757.1 NM_032844.4 NR_135469.1 XM_005252631.3 XM_005252632.3 XM_006717519.3 XM_011519751.2 XM_017016852.1 XM_017016853.1 XM_017016856.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
48555 c.2560G>A p.D854N Chr.10: 27186459-27186459 on Build GRCh38
5654152 c.2616T>C p.T872T Chr.10: 27186515-27186515 on Build GRCh38
7284854 c.2542C>T p.P848S Chr.10: 27186441-27186441 on Build GRCh38
7178308 c.2624G>A p.G875E Chr.10: 27186523-27186523 on Build GRCh38
7152488 c.2563G>A p.D855N Chr.10: 27186462-27186462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201251613 Chr.10: 27186544-27186544 on Build GRCh38
rs763021387 Chr.10: 27186523-27186523 on Build GRCh38
rs570139166 Chr.10: 27186596-27186596 on Build GRCh38
rs757986015 Chr.10: 27186427-27186427 on Build GRCh38
rs886046961 Chr.10: 27186422-27186422 on Build GRCh38
rs1030341538 Chr.10: 27186595-27186595 on Build GRCh38
rs886046960 Chr.10: 27186413-27186413 on Build GRCh38
rs377115842 Chr.10: 27186582-27186582 on Build GRCh38
rs779790086 Chr.10: 27186462-27186462 on Build GRCh38
rs199527702 Chr.10: 27186566-27186566 on Build GRCh38
rs768523040 Chr.10: 27186499-27186499 on Build GRCh38
rs756568425 Chr.10: 27186572-27186572 on Build GRCh38
rs764732210 Chr.10: 27186578-27186578 on Build GRCh38
rs746673440 Chr.10: 27186491-27186491 on Build GRCh38
rs138288481 Chr.10: 27186516-27186516 on Build GRCh38
rs10741130 Chr.10: 27186515-27186515 on Build GRCh38
rs774504350 Chr.10: 27186563-27186563 on Build GRCh38
rs375733380 Chr.10: 27186424-27186424 on Build GRCh38
rs10764686 Chr.10: 27186565-27186565 on Build GRCh38
rs748005651 Chr.10: 27186512-27186512 on Build GRCh38
rs902207437 Chr.10: 27186567-27186568 on Build GRCh38
rs568543576 Chr.10: 27186562-27186562 on Build GRCh38
rs752083792 Chr.10: 27186587-27186587 on Build GRCh38
rs369916275 Chr.10: 27186504-27186504 on Build GRCh38
rs200773075 Chr.10: 27186470-27186470 on Build GRCh38
rs778267080 Chr.10: 27186420-27186420 on Build GRCh38

View Full Product Details