Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75932906-75933139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO22
Entrez Gene ID
26263
Gene Name
F-box protein 22
Gene Aliases
FBX22, FISTC1
UniGene
Hs.591115
Gene Chromosome Location
Chr.15: 75903859-75937513 on Build GRCh38
Transcripts
NM_147188.2 NR_037623.1 XM_011521447.2 NR_003136.1

Gene Symbol
FBXO22-AS1
Entrez Gene ID
692224
Gene Name
FBXO22 antisense RNA 1
Gene Aliases
FBXO22OS, FISTC1OS, HsT18082, NCRNA00053
UniGene
Hs.591115
Gene Chromosome Location
Chr.15: 75932683-75933130 on Build GRCh38
Transcripts
NM_147188.2 NR_037623.1 XM_011521447.2 NR_003136.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
416807 c.1060T>G p.L354V Chr.15: 75932950-75932950 on Build GRCh38
3504153 c.1079A>C p.N360T Chr.15: 75932969-75932969 on Build GRCh38
5470400 c.1098T>A p.D366E Chr.15: 75932988-75932988 on Build GRCh38
701822 c.1208A>G p.K403R Chr.15: 75933098-75933098 on Build GRCh38
965087 c.1086A>C p.E362D Chr.15: 75932976-75932976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765931064 Chr.15: 75933098-75933100 on Build GRCh38
rs771825258 Chr.15: 75933048-75933048 on Build GRCh38
rs267604328 Chr.15: 75932973-75932973 on Build GRCh38
rs150074021 Chr.15: 75932950-75932950 on Build GRCh38
rs773806650 Chr.15: 75933037-75933039 on Build GRCh38
rs991709953 Chr.15: 75933099-75933099 on Build GRCh38
rs945582178 Chr.15: 75933089-75933089 on Build GRCh38
rs773405572 Chr.15: 75932941-75932941 on Build GRCh38
rs753312397 Chr.15: 75932976-75932976 on Build GRCh38
rs113146835 Chr.15: 75933066-75933066 on Build GRCh38
rs748679911 Chr.15: 75933012-75933012 on Build GRCh38
rs774350053 Chr.15: 75932933-75932933 on Build GRCh38
rs759516928 Chr.15: 75933079-75933079 on Build GRCh38
rs758812767 Chr.15: 75932979-75932979 on Build GRCh38
rs34255722 Chr.15: 75932958-75932958 on Build GRCh38
rs770473372 Chr.15: 75933006-75933007 on Build GRCh38
rs755088177 Chr.15: 75932956-75932956 on Build GRCh38
rs958919463 Chr.15: 75933073-75933073 on Build GRCh38
rs778706225 Chr.15: 75933018-75933018 on Build GRCh38
rs773901647 Chr.15: 75933103-75933106 on Build GRCh38
rs767361655 Chr.15: 75932939-75932939 on Build GRCh38
rs567773739 Chr.15: 75932940-75932940 on Build GRCh38
rs995439148 Chr.15: 75933064-75933064 on Build GRCh38
rs762558195 Chr.15: 75933096-75933099 on Build GRCh38
rs1015124403 Chr.15: 75932955-75932955 on Build GRCh38
rs768280681 Chr.15: 75933013-75933013 on Build GRCh38
rs776429552 Chr.15: 75933067-75933067 on Build GRCh38
rs766657640 Chr.15: 75932949-75932949 on Build GRCh38
rs770989647 Chr.15: 75933070-75933070 on Build GRCh38
rs536714391 Chr.15: 75933055-75933055 on Build GRCh38
rs143665207 Chr.15: 75933102-75933102 on Build GRCh38
rs761593704 Chr.15: 75932934-75932934 on Build GRCh38
rs748135980 Chr.15: 75933030-75933030 on Build GRCh38
rs760929745 Chr.15: 75932943-75932943 on Build GRCh38
rs776975292 Chr.15: 75933078-75933078 on Build GRCh38
rs755780709 Chr.15: 75933001-75933001 on Build GRCh38
rs760473711 Chr.15: 75933059-75933059 on Build GRCh38
rs753799726 Chr.15: 75932953-75932953 on Build GRCh38
rs141220458 Chr.15: 75932990-75932990 on Build GRCh38
rs779772186 Chr.15: 75933011-75933011 on Build GRCh38

View Full Product Details