Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 71215615-71215848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EYA1
Entrez Gene ID
2138
Gene Name
EYA transcriptional coactivator and phosphatase 1
Gene Aliases
BOP, BOR, BOS1, OFC1
UniGene
Hs.444971
Gene Chromosome Location
Chr.8: 71197433-71548130 on Build GRCh38
Transcripts
NM_000503.5 NM_001288574.1 NM_001288575.1 NM_172058.3 NM_172059.3 NM_172060.3 XM_011517483.2 XM_011517484.2 XM_017013201.1 XM_017013202.1 XM_017013203.1 XM_017013204.1 XM_017013205.1 XM_017013206.1 XM_017013207.1 XM_017013208.1 XM_017013209.1 XM_017013210.1 XM_017013211.1 XM_017013212.1 XM_017013213.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6181618 c.1417G>T p.A473S Chr.8: 71215672-71215672 on Build GRCh38
4953991 c.1371T>A p.G457G Chr.8: 71215718-71215718 on Build GRCh38
5912800 c.1307C>T p.A436V Chr.8: 71215683-71215683 on Build GRCh38
6657454 c.1420C>A p.L474M Chr.8: 71215669-71215669 on Build GRCh38
397899 c.1434G>T p.W478C Chr.8: 71215655-71215655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781247880 Chr.8: 71215688-71215688 on Build GRCh38
rs112593082 Chr.8: 71215712-71215712 on Build GRCh38
rs758650723 Chr.8: 71215670-71215670 on Build GRCh38
rs767194310 Chr.8: 71215747-71215747 on Build GRCh38
rs763898906 Chr.8: 71215772-71215772 on Build GRCh38
rs778970223 Chr.8: 71215635-71215635 on Build GRCh38
rs767104377 Chr.8: 71215648-71215648 on Build GRCh38
rs540131742 Chr.8: 71215706-71215706 on Build GRCh38
rs774803914 Chr.8: 71215762-71215762 on Build GRCh38
rs755991512 Chr.8: 71215659-71215659 on Build GRCh38
rs760666558 Chr.8: 71215771-71215771 on Build GRCh38
rs980029055 Chr.8: 71215787-71215787 on Build GRCh38
rs375836302 Chr.8: 71215724-71215724 on Build GRCh38
rs771397353 Chr.8: 71215717-71215717 on Build GRCh38
rs752745332 Chr.8: 71215654-71215654 on Build GRCh38
rs780203392 Chr.8: 71215681-71215681 on Build GRCh38
rs201078425 Chr.8: 71215682-71215682 on Build GRCh38
rs189471908 Chr.8: 71215781-71215781 on Build GRCh38
rs727503044 Chr.8: 71215687-71215687 on Build GRCh38
rs397517917 Chr.8: 71215729-71215729 on Build GRCh38
rs770129052 Chr.8: 71215705-71215705 on Build GRCh38
rs727503043 Chr.8: 71215664-71215664 on Build GRCh38
rs755093166 Chr.8: 71215686-71215686 on Build GRCh38
rs371408686 Chr.8: 71215663-71215663 on Build GRCh38
rs55890504 Chr.8: 71215738-71215738 on Build GRCh38
rs759255995 Chr.8: 71215743-71215743 on Build GRCh38
rs1021483963 Chr.8: 71215796-71215796 on Build GRCh38
rs748750618 Chr.8: 71215690-71215690 on Build GRCh38
rs772827985 Chr.8: 71215780-71215780 on Build GRCh38
rs200995253 Chr.8: 71215788-71215788 on Build GRCh38
rs997974243 Chr.8: 71215703-71215703 on Build GRCh38

View Full Product Details