Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 38905641-38905914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM114A1
Entrez Gene ID
92689
Gene Name
family with sequence similarity 114 member A1
Gene Aliases
Noxp20
UniGene
Hs.476517
Gene Chromosome Location
Chr.4: 38867733-38945744 on Build GRCh38
Transcripts
NM_138389.2 NR_033290.1 XM_005262672.2 XM_006714033.2 XM_017008838.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1429581 c.590C>T p.P197L Chr.4: 38905794-38905794 on Build GRCh38
6100028 c.580G>T p.A194S Chr.4: 38905784-38905784 on Build GRCh38
6658108 c.617G>A p.R206Q Chr.4: 38905821-38905821 on Build GRCh38
4881042 c.657+10T>C p.? Chr.4: 38905871-38905871 on Build GRCh38
733524 c.617G>T p.R206L Chr.4: 38905821-38905821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761221211 Chr.4: 38905733-38905733 on Build GRCh38
rs760885494 Chr.4: 38905666-38905666 on Build GRCh38
rs906021746 Chr.4: 38905702-38905702 on Build GRCh38
rs372801338 Chr.4: 38905869-38905869 on Build GRCh38
rs763959361 Chr.4: 38905678-38905678 on Build GRCh38
rs1055208615 Chr.4: 38905736-38905736 on Build GRCh38
rs777277396 Chr.4: 38905861-38905861 on Build GRCh38
rs1027418475 Chr.4: 38905815-38905815 on Build GRCh38
rs775617833 Chr.4: 38905870-38905870 on Build GRCh38
rs747803554 Chr.4: 38905816-38905816 on Build GRCh38
rs139238894 Chr.4: 38905784-38905784 on Build GRCh38
rs754142280 Chr.4: 38905675-38905675 on Build GRCh38
rs139789863 Chr.4: 38905820-38905820 on Build GRCh38
rs763196324 Chr.4: 38905877-38905877 on Build GRCh38
rs773250651 Chr.4: 38905710-38905710 on Build GRCh38
rs752953862 Chr.4: 38905765-38905765 on Build GRCh38
rs778231131 Chr.4: 38905821-38905821 on Build GRCh38
rs937615570 Chr.4: 38905705-38905705 on Build GRCh38
rs187220962 Chr.4: 38905663-38905663 on Build GRCh38
rs554734104 Chr.4: 38905735-38905735 on Build GRCh38
rs766584561 Chr.4: 38905739-38905739 on Build GRCh38
rs748518530 Chr.4: 38905718-38905718 on Build GRCh38
rs751249984 Chr.4: 38905679-38905679 on Build GRCh38
rs755103652 Chr.4: 38905677-38905677 on Build GRCh38
rs372382288 Chr.4: 38905754-38905754 on Build GRCh38
rs750392881 Chr.4: 38905780-38905780 on Build GRCh38
rs976663662 Chr.4: 38905823-38905823 on Build GRCh38
rs772457949 Chr.4: 38905720-38905720 on Build GRCh38
rs144537370 Chr.4: 38905778-38905778 on Build GRCh38
rs747527647 Chr.4: 38905824-38905824 on Build GRCh38
rs200345622 Chr.4: 38905776-38905776 on Build GRCh38
rs760610875 Chr.4: 38905892-38905892 on Build GRCh38
rs430296 Chr.4: 38905806-38905806 on Build GRCh38
rs750161596 Chr.4: 38905889-38905889 on Build GRCh38
rs543114574 Chr.4: 38905768-38905768 on Build GRCh38
rs776790371 Chr.4: 38905753-38905753 on Build GRCh38
rs765676345 Chr.4: 38905713-38905713 on Build GRCh38
rs1029962015 Chr.4: 38905843-38905843 on Build GRCh38
rs199533608 Chr.4: 38905670-38905670 on Build GRCh38
rs779553902 Chr.4: 38905785-38905785 on Build GRCh38
rs199977746 Chr.4: 38905868-38905868 on Build GRCh38
rs767324729 Chr.4: 38905879-38905879 on Build GRCh38
rs781168047 Chr.4: 38905683-38905683 on Build GRCh38
rs376696883 Chr.4: 38905681-38905681 on Build GRCh38
rs965243657 Chr.4: 38905835-38905835 on Build GRCh38
rs111831236 Chr.4: 38905721-38905721 on Build GRCh38
rs765447912 Chr.4: 38905764-38905764 on Build GRCh38
rs755662903 Chr.4: 38905685-38905685 on Build GRCh38
rs1018110269 Chr.4: 38905826-38905826 on Build GRCh38
rs142423470 Chr.4: 38905786-38905786 on Build GRCh38
rs774048175 Chr.4: 38905717-38905717 on Build GRCh38
rs754497110 Chr.4: 38905790-38905790 on Build GRCh38
rs768136264 Chr.4: 38905716-38905716 on Build GRCh38
rs745785618 Chr.4: 38905684-38905684 on Build GRCh38

View Full Product Details