Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 4759480-4759747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXK1
Entrez Gene ID
221937
Gene Name
forkhead box K1
Gene Aliases
FOXK1L
UniGene
Hs.487393
Gene Chromosome Location
Chr.7: 4644691-4771443 on Build GRCh38
Transcripts
NM_001037165.1 XM_011515191.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7189602 c.1638G>A p.A546A Chr.7: 4759537-4759537 on Build GRCh38
4631210 c.1568C>T p.A523V Chr.7: 4759533-4759533 on Build GRCh38
5568550 c.1581C>G p.S527S Chr.7: 4759546-4759546 on Build GRCh38
5514190 c.1633G>A p.A545T Chr.7: 4759532-4759532 on Build GRCh38
3762700 c.1611A>T p.G537G Chr.7: 4759510-4759510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005380627 Chr.7: 4759546-4759546 on Build GRCh38
rs764273089 Chr.7: 4759512-4759512 on Build GRCh38
rs754783299 Chr.7: 4759534-4759534 on Build GRCh38
rs751569462 Chr.7: 4759514-4759514 on Build GRCh38
rs950365148 Chr.7: 4759654-4759654 on Build GRCh38
rs368805899 Chr.7: 4759585-4759585 on Build GRCh38
rs370502359 Chr.7: 4759555-4759555 on Build GRCh38
rs778526385 Chr.7: 4759536-4759536 on Build GRCh38
rs1003844502 Chr.7: 4759533-4759533 on Build GRCh38
rs370912425 Chr.7: 4759523-4759523 on Build GRCh38
rs955536700 Chr.7: 4759633-4759634 on Build GRCh38
rs574608935 Chr.7: 4759646-4759646 on Build GRCh38
rs895060594 Chr.7: 4759699-4759699 on Build GRCh38
rs374135989 Chr.7: 4759548-4759548 on Build GRCh38
rs757334416 Chr.7: 4759610-4759610 on Build GRCh38
rs564566124 Chr.7: 4759681-4759681 on Build GRCh38
rs1007087610 Chr.7: 4759635-4759635 on Build GRCh38
rs897004581 Chr.7: 4759708-4759708 on Build GRCh38
rs988089378 Chr.7: 4759578-4759578 on Build GRCh38
rs772831526 Chr.7: 4759544-4759544 on Build GRCh38
rs752984376 Chr.7: 4759508-4759508 on Build GRCh38
rs560240503 Chr.7: 4759650-4759650 on Build GRCh38
rs367605292 Chr.7: 4759558-4759558 on Build GRCh38
rs906485216 Chr.7: 4759675-4759675 on Build GRCh38
rs542129332 Chr.7: 4759649-4759649 on Build GRCh38
rs544000290 Chr.7: 4759530-4759530 on Build GRCh38
rs558240328 Chr.7: 4759504-4759504 on Build GRCh38
rs945433282 Chr.7: 4759666-4759666 on Build GRCh38
rs1055569612 Chr.7: 4759687-4759687 on Build GRCh38
rs1029416348 Chr.7: 4759723-4759723 on Build GRCh38
rs377464608 Chr.7: 4759516-4759516 on Build GRCh38
rs118117357 Chr.7: 4759701-4759701 on Build GRCh38
rs370318084 Chr.7: 4759567-4759567 on Build GRCh38
rs550380974 Chr.7: 4759717-4759717 on Build GRCh38
rs972506783 Chr.7: 4759561-4759561 on Build GRCh38
rs374908293 Chr.7: 4759557-4759557 on Build GRCh38
rs527694290 Chr.7: 4759651-4759651 on Build GRCh38
rs764208323 Chr.7: 4759590-4759590 on Build GRCh38
rs770495931 Chr.7: 4759552-4759552 on Build GRCh38
rs866678847 Chr.7: 4759554-4759554 on Build GRCh38
rs983179523 Chr.7: 4759662-4759662 on Build GRCh38
rs374700671 Chr.7: 4759568-4759568 on Build GRCh38
rs1029728126 Chr.7: 4759623-4759623 on Build GRCh38
rs11980294 Chr.7: 4759669-4759669 on Build GRCh38
rs943660281 Chr.7: 4759627-4759627 on Build GRCh38
rs917027495 Chr.7: 4759668-4759668 on Build GRCh38
rs767529198 Chr.7: 4759622-4759622 on Build GRCh38
rs79357656 Chr.7: 4759510-4759510 on Build GRCh38
rs200818519 Chr.7: 4759540-4759540 on Build GRCh38
rs751728155 Chr.7: 4759608-4759608 on Build GRCh38
rs765637261 Chr.7: 4759507-4759507 on Build GRCh38
rs989607310 Chr.7: 4759628-4759628 on Build GRCh38
rs747868947 Chr.7: 4759539-4759539 on Build GRCh38
rs1016765061 Chr.7: 4759702-4759702 on Build GRCh38
rs763284610 Chr.7: 4759586-4759586 on Build GRCh38
rs759980175 Chr.7: 4759506-4759506 on Build GRCh38

View Full Product Details