Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 31369340-31369605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FABP3
Entrez Gene ID
2170
Gene Name
fatty acid binding protein 3
Gene Aliases
FABP11, H-FABP, M-FABP, MDGI, O-FABP
UniGene
Hs.576372
Gene Chromosome Location
Chr.1: 31359611-31373618 on Build GRCh38
Transcripts
NM_001320996.1 NM_004102.4 XM_011541007.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6450269 c.120C>T p.T40T Chr.1: 31369511-31369511 on Build GRCh38
6357727 c.194T>G p.F65C Chr.1: 31369437-31369437 on Build GRCh38
6320330 c.144C>A p.D48E Chr.1: 31369487-31369487 on Build GRCh38
3804925 c.206T>G p.V69G Chr.1: 31369425-31369425 on Build GRCh38
7126724 c.158A>G p.K53R Chr.1: 31369473-31369473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771606487 Chr.1: 31369407-31369407 on Build GRCh38
rs766625223 Chr.1: 31369444-31369444 on Build GRCh38
rs200903923 Chr.1: 31369448-31369448 on Build GRCh38
rs938624847 Chr.1: 31369571-31369571 on Build GRCh38
rs113138316 Chr.1: 31369384-31369384 on Build GRCh38
rs200478203 Chr.1: 31369373-31369373 on Build GRCh38
rs774014193 Chr.1: 31369385-31369385 on Build GRCh38
rs866763179 Chr.1: 31369525-31369525 on Build GRCh38
rs770672392 Chr.1: 31369375-31369375 on Build GRCh38
rs901514519 Chr.1: 31369458-31369458 on Build GRCh38
rs762380126 Chr.1: 31369548-31369548 on Build GRCh38
rs113861239 Chr.1: 31369559-31369559 on Build GRCh38
rs774940953 Chr.1: 31369512-31369512 on Build GRCh38
rs200702682 Chr.1: 31369408-31369408 on Build GRCh38
rs146114949 Chr.1: 31369502-31369502 on Build GRCh38
rs945785374 Chr.1: 31369438-31369438 on Build GRCh38
rs768005655 Chr.1: 31369527-31369527 on Build GRCh38
rs765678476 Chr.1: 31369431-31369431 on Build GRCh38
rs745493497 Chr.1: 31369393-31369393 on Build GRCh38
rs758689942 Chr.1: 31369441-31369441 on Build GRCh38
rs201441558 Chr.1: 31369531-31369531 on Build GRCh38
rs764669863 Chr.1: 31369418-31369418 on Build GRCh38
rs779315102 Chr.1: 31369494-31369496 on Build GRCh38
rs750714301 Chr.1: 31369435-31369435 on Build GRCh38
rs770350802 Chr.1: 31369555-31369555 on Build GRCh38
rs746276453 Chr.1: 31369526-31369529 on Build GRCh38
rs751675000 Chr.1: 31369461-31369461 on Build GRCh38
rs746563492 Chr.1: 31369524-31369524 on Build GRCh38
rs907142744 Chr.1: 31369541-31369541 on Build GRCh38
rs749130570 Chr.1: 31369490-31369490 on Build GRCh38
rs201934570 Chr.1: 31369508-31369508 on Build GRCh38
rs2228194 Chr.1: 31369473-31369473 on Build GRCh38
rs771696636 Chr.1: 31369510-31369510 on Build GRCh38
rs200154349 Chr.1: 31369446-31369446 on Build GRCh38
rs200003616 Chr.1: 31369507-31369507 on Build GRCh38
rs757091758 Chr.1: 31369501-31369501 on Build GRCh38
rs111898472 Chr.1: 31369383-31369383 on Build GRCh38
rs372734842 Chr.1: 31369560-31369560 on Build GRCh38
rs776966330 Chr.1: 31369421-31369421 on Build GRCh38
rs376655464 Chr.1: 31369575-31369575 on Build GRCh38
rs369006737 Chr.1: 31369417-31369417 on Build GRCh38
rs762294497 Chr.1: 31369429-31369429 on Build GRCh38
rs1040123276 Chr.1: 31369448-31369449 on Build GRCh38
rs483352874 Chr.1: 31369368-31369369 on Build GRCh38

View Full Product Details