Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 435574-435807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO25
Entrez Gene ID
26260
Gene Name
F-box protein 25
Gene Aliases
FBX25
UniGene
Hs.438454
Gene Chromosome Location
Chr.8: 406808-477967 on Build GRCh38
Transcripts
NM_012173.3 NM_183420.1 NM_183421.1 XM_011534748.2 XM_011534749.1 XM_017013313.1 XM_017013314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1099665 c.315A>G p.E105E Chr.8: 435641-435641 on Build GRCh38
3648934 c.357C>T p.I119I Chr.8: 435683-435683 on Build GRCh38
4965304 c.376G>A p.V126I Chr.8: 435702-435702 on Build GRCh38
4489589 c.349C>T p.Q117* Chr.8: 435675-435675 on Build GRCh38
3834691 c.357C>A p.I119I Chr.8: 435683-435683 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201221465 Chr.8: 435717-435717 on Build GRCh38
rs371366010 Chr.8: 435665-435665 on Build GRCh38
rs561294577 Chr.8: 435739-435739 on Build GRCh38
rs777406076 Chr.8: 435644-435644 on Build GRCh38
rs71514155 Chr.8: 435786-435786 on Build GRCh38
rs369367871 Chr.8: 435675-435675 on Build GRCh38
rs745860388 Chr.8: 435714-435715 on Build GRCh38
rs140242296 Chr.8: 435664-435664 on Build GRCh38
rs765177275 Chr.8: 435684-435684 on Build GRCh38
rs17665428 Chr.8: 435612-435612 on Build GRCh38
rs147533889 Chr.8: 435652-435652 on Build GRCh38
rs547775550 Chr.8: 435732-435732 on Build GRCh38
rs565516107 Chr.8: 435712-435712 on Build GRCh38
rs756126393 Chr.8: 435730-435730 on Build GRCh38
rs768397227 Chr.8: 435671-435671 on Build GRCh38
rs752818460 Chr.8: 435619-435619 on Build GRCh38
rs769616878 Chr.8: 435751-435751 on Build GRCh38
rs745757008 Chr.8: 435725-435725 on Build GRCh38
rs778117976 Chr.8: 435698-435698 on Build GRCh38
rs767057643 Chr.8: 435611-435611 on Build GRCh38
rs576684461 Chr.8: 435680-435680 on Build GRCh38
rs781053280 Chr.8: 435657-435657 on Build GRCh38
rs758515533 Chr.8: 435691-435691 on Build GRCh38
rs758665822 Chr.8: 435625-435625 on Build GRCh38
rs754328792 Chr.8: 435682-435682 on Build GRCh38
rs748897794 Chr.8: 435735-435735 on Build GRCh38
rs914815426 Chr.8: 435775-435775 on Build GRCh38
rs761178308 Chr.8: 435677-435677 on Build GRCh38
rs779054336 Chr.8: 435617-435617 on Build GRCh38
rs747803249 Chr.8: 435756-435756 on Build GRCh38
rs769150294 Chr.8: 435666-435666 on Build GRCh38
rs755049263 Chr.8: 435614-435614 on Build GRCh38
rs202007568 Chr.8: 435629-435629 on Build GRCh38
rs781000218 Chr.8: 435721-435721 on Build GRCh38
rs71514154 Chr.8: 435608-435608 on Build GRCh38
rs530055479 Chr.8: 435742-435742 on Build GRCh38
rs199702170 Chr.8: 435685-435685 on Build GRCh38
rs774086353 Chr.8: 435754-435754 on Build GRCh38
rs200299799 Chr.8: 435667-435667 on Build GRCh38
rs760120607 Chr.8: 435683-435683 on Build GRCh38

View Full Product Details