Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 173303965-173304198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT7
Entrez Gene ID
51809
Gene Name
polypeptide N-acetylgalactosaminyltransferase 7
Gene Aliases
GALNAC-T7, GalNAcT7
UniGene
Hs.548088
Gene Chromosome Location
Chr.4: 173168753-173323967 on Build GRCh38
Transcripts
NM_017423.2 XM_005263062.1 XM_011532025.2 XM_017008292.1 XM_017008293.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6027040 c.1352C>A p.P451H Chr.4: 173304081-173304081 on Build GRCh38
3993548 c.1267-1G>A p.? Chr.4: 173303995-173303995 on Build GRCh38
3602051 c.1300C>T p.P434S Chr.4: 173304029-173304029 on Build GRCh38
332950 c.1373C>G p.S458C Chr.4: 173304102-173304102 on Build GRCh38
6669122 c.1356G>A p.P452P Chr.4: 173304085-173304085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753416672 Chr.4: 173304158-173304161 on Build GRCh38
rs755410918 Chr.4: 173304051-173304051 on Build GRCh38
rs573676611 Chr.4: 173304134-173304134 on Build GRCh38
rs771857869 Chr.4: 173304153-173304153 on Build GRCh38
rs774118713 Chr.4: 173304126-173304126 on Build GRCh38
rs766814544 Chr.4: 173304032-173304032 on Build GRCh38
rs770637602 Chr.4: 173304089-173304089 on Build GRCh38
rs763578569 Chr.4: 173304123-173304124 on Build GRCh38
rs775455913 Chr.4: 173304164-173304164 on Build GRCh38
rs781738624 Chr.4: 173304056-173304056 on Build GRCh38
rs144428597 Chr.4: 173304010-173304010 on Build GRCh38
rs765261051 Chr.4: 173303992-173303992 on Build GRCh38
rs948709638 Chr.4: 173304118-173304118 on Build GRCh38
rs758594665 Chr.4: 173304024-173304024 on Build GRCh38
rs920013656 Chr.4: 173304108-173304108 on Build GRCh38
rs777199234 Chr.4: 173304084-173304084 on Build GRCh38
rs755603920 Chr.4: 173304082-173304082 on Build GRCh38
rs765141870 Chr.4: 173304163-173304163 on Build GRCh38
rs377071584 Chr.4: 173304095-173304095 on Build GRCh38
rs143154840 Chr.4: 173303996-173303996 on Build GRCh38
rs559934063 Chr.4: 173304039-173304039 on Build GRCh38
rs150824797 Chr.4: 173304085-173304085 on Build GRCh38
rs748636968 Chr.4: 173304057-173304057 on Build GRCh38

View Full Product Details