Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 135941058-135941303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL21
Entrez Gene ID
26223
Gene Name
F-box and leucine rich repeat protein 21 (gene/pseudogene)
Gene Aliases
FBL3B, FBXL3B, FBXL3P, Fbl21
UniGene
Hs.591275
Gene Chromosome Location
Chr.5: 135930317-135941678 on Build GRCh38
Transcripts
NM_012159.4 NR_132347.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5918643 c.376G>A p.D126N Chr.5: 135941216-135941216 on Build GRCh38
5987712 c.901C>G p.L301V Chr.5: 135941267-135941267 on Build GRCh38
275138 c.444C>T p.F148F Chr.5: 135941284-135941284 on Build GRCh38
4407328 c.759G>A p.E253E Chr.5: 135941125-135941125 on Build GRCh38
260896 c.463G>A p.E155K Chr.5: 135941303-135941303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201988376 Chr.5: 135941194-135941194 on Build GRCh38
rs746835230 Chr.5: 135941123-135941123 on Build GRCh38
rs767833646 Chr.5: 135941080-135941080 on Build GRCh38
rs752778751 Chr.5: 135941192-135941192 on Build GRCh38
rs765141607 Chr.5: 135941113-135941113 on Build GRCh38
rs373193586 Chr.5: 135941100-135941100 on Build GRCh38
rs768407641 Chr.5: 135941152-135941152 on Build GRCh38
rs1034852031 Chr.5: 135941273-135941273 on Build GRCh38
rs1051053081 Chr.5: 135941269-135941269 on Build GRCh38
rs760748094 Chr.5: 135941094-135941094 on Build GRCh38
rs780936197 Chr.5: 135941138-135941138 on Build GRCh38
rs565728948 Chr.5: 135941107-135941107 on Build GRCh38
rs535648953 Chr.5: 135941228-135941228 on Build GRCh38
rs935263193 Chr.5: 135941211-135941211 on Build GRCh38
rs376112540 Chr.5: 135941110-135941110 on Build GRCh38
rs769485830 Chr.5: 135941145-135941145 on Build GRCh38
rs2066913 Chr.5: 135941125-135941125 on Build GRCh38
rs779649153 Chr.5: 135941276-135941276 on Build GRCh38
rs370773703 Chr.5: 135941122-135941122 on Build GRCh38
rs780978283 Chr.5: 135941237-135941237 on Build GRCh38
rs916371096 Chr.5: 135941183-135941183 on Build GRCh38
rs946519357 Chr.5: 135941186-135941186 on Build GRCh38
rs759602930 Chr.5: 135941166-135941166 on Build GRCh38
rs773626132 Chr.5: 135941082-135941082 on Build GRCh38
rs750166472 Chr.5: 135941261-135941261 on Build GRCh38
rs765368729 Chr.5: 135941180-135941180 on Build GRCh38
rs772643518 Chr.5: 135941151-135941151 on Build GRCh38
rs766481344 Chr.5: 135941159-135941159 on Build GRCh38
rs748594631 Chr.5: 135941150-135941150 on Build GRCh38
rs757123098 Chr.5: 135941225-135941225 on Build GRCh38
rs549266512 Chr.5: 135941228-135941228 on Build GRCh38
rs774990449 Chr.5: 135941147-135941147 on Build GRCh38
rs746613480 Chr.5: 135941134-135941134 on Build GRCh38
rs755829763 Chr.5: 135941270-135941270 on Build GRCh38
rs758208952 Chr.5: 135941116-135941116 on Build GRCh38
rs986290045 Chr.5: 135941114-135941114 on Build GRCh38
rs31547 Chr.5: 135941158-135941158 on Build GRCh38
rs764115822 Chr.5: 135941193-135941193 on Build GRCh38

View Full Product Details