Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73227096-73227327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM104A
Entrez Gene ID
84923
Gene Name
family with sequence similarity 104 member A
Gene Aliases
-
UniGene
Hs.741276
Gene Chromosome Location
Chr.17: 73207353-73232394 on Build GRCh38
Transcripts
NM_001098832.1 NM_001289410.1 NM_001289411.1 NM_001289412.1 NM_032837.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6658034 c.83G>C p.S28T Chr.17: 73227205-73227205 on Build GRCh38
6658036 c.45C>T p.N15N Chr.17: 73227243-73227243 on Build GRCh38
6319032 c.301C>A p.Q101K Chr.17: 73227185-73227185 on Build GRCh38
7303264 c.80G>T p.R27I Chr.17: 73227208-73227208 on Build GRCh38
1251692 c.70delC p.Q24fs*48 Chr.17: 73227218-73227218 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755556056 Chr.17: 73227200-73227200 on Build GRCh38
rs776261735 Chr.17: 73227277-73227277 on Build GRCh38
rs768397322 Chr.17: 73227232-73227232 on Build GRCh38
rs150198925 Chr.17: 73227129-73227129 on Build GRCh38
rs185062077 Chr.17: 73227157-73227157 on Build GRCh38
rs759837866 Chr.17: 73227270-73227270 on Build GRCh38
rs4550516 Chr.17: 73227237-73227237 on Build GRCh38
rs148088779 Chr.17: 73227234-73227234 on Build GRCh38
rs763681430 Chr.17: 73227288-73227288 on Build GRCh38
rs756906474 Chr.17: 73227300-73227300 on Build GRCh38
rs71154980 Chr.17: 73227286-73227286 on Build GRCh38
rs75193968 Chr.17: 73227297-73227298 on Build GRCh38
rs370741544 Chr.17: 73227219-73227219 on Build GRCh38
rs775315535 Chr.17: 73227224-73227224 on Build GRCh38
rs752201911 Chr.17: 73227197-73227197 on Build GRCh38
rs759234405 Chr.17: 73227285-73227286 on Build GRCh38
rs866843450 Chr.17: 73227242-73227242 on Build GRCh38
rs980534628 Chr.17: 73227264-73227264 on Build GRCh38
rs750931255 Chr.17: 73227298-73227299 on Build GRCh38
rs766603126 Chr.17: 73227243-73227243 on Build GRCh38
rs753112846 Chr.17: 73227281-73227281 on Build GRCh38
rs748940049 Chr.17: 73227218-73227218 on Build GRCh38
rs368047723 Chr.17: 73227298-73227298 on Build GRCh38
rs182003333 Chr.17: 73227127-73227127 on Build GRCh38
rs773297769 Chr.17: 73227283-73227284 on Build GRCh38
rs751185974 Chr.17: 73227139-73227140 on Build GRCh38
rs769947396 Chr.17: 73227217-73227218 on Build GRCh38
rs372168359 Chr.17: 73227285-73227285 on Build GRCh38
rs763150088 Chr.17: 73227240-73227240 on Build GRCh38
rs755445865 Chr.17: 73227134-73227134 on Build GRCh38
rs777461616 Chr.17: 73227214-73227214 on Build GRCh38
rs753064534 Chr.17: 73227287-73227287 on Build GRCh38
rs767974174 Chr.17: 73227276-73227276 on Build GRCh38
rs373649856 Chr.17: 73227225-73227225 on Build GRCh38
rs752308696 Chr.17: 73227286-73227288 on Build GRCh38
rs367850869 Chr.17: 73227220-73227220 on Build GRCh38
rs77933932 Chr.17: 73227286-73227286 on Build GRCh38
rs145613050 Chr.17: 73227223-73227223 on Build GRCh38
rs774735936 Chr.17: 73227245-73227245 on Build GRCh38
rs767329194 Chr.17: 73227286-73227287 on Build GRCh38

View Full Product Details