Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 6535997-6536270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLDC
Entrez Gene ID
2731
Gene Name
glycine decarboxylase
Gene Aliases
GCE, GCSP, HYGN1
UniGene
Hs.584238
Gene Chromosome Location
Chr.9: 6532464-6645692 on Build GRCh38
Transcripts
NM_000170.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3367737 c.2683A>G p.M895V Chr.9: 6536219-6536219 on Build GRCh38
1285167 c.2797A>T p.I933F Chr.9: 6536105-6536105 on Build GRCh38
3907608 c.2831C>T p.P944L Chr.9: 6536071-6536071 on Build GRCh38
6861563 c.2705C>T p.T902I Chr.9: 6536197-6536197 on Build GRCh38
6183600 c.2711T>C p.M904T Chr.9: 6536191-6536191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766928717 Chr.9: 6536093-6536093 on Build GRCh38
rs1039258333 Chr.9: 6536250-6536250 on Build GRCh38
rs928868906 Chr.9: 6536045-6536045 on Build GRCh38
rs772682541 Chr.9: 6536055-6536055 on Build GRCh38
rs771664814 Chr.9: 6536043-6536043 on Build GRCh38
rs374022098 Chr.9: 6536160-6536160 on Build GRCh38
rs747879174 Chr.9: 6536103-6536103 on Build GRCh38
rs371186899 Chr.9: 6536067-6536067 on Build GRCh38
rs151100082 Chr.9: 6536092-6536092 on Build GRCh38
rs144937031 Chr.9: 6536172-6536172 on Build GRCh38
rs188269735 Chr.9: 6536188-6536188 on Build GRCh38
rs759933539 Chr.9: 6536173-6536173 on Build GRCh38
rs773430894 Chr.9: 6536156-6536156 on Build GRCh38
rs1007363577 Chr.9: 6536240-6536240 on Build GRCh38
rs542056100 Chr.9: 6536063-6536063 on Build GRCh38
rs758029533 Chr.9: 6536104-6536104 on Build GRCh38
rs776573369 Chr.9: 6536245-6536245 on Build GRCh38
rs758046295 Chr.9: 6536198-6536198 on Build GRCh38
rs769670556 Chr.9: 6536143-6536143 on Build GRCh38
rs779129201 Chr.9: 6536101-6536101 on Build GRCh38
rs745769719 Chr.9: 6536129-6536129 on Build GRCh38
rs139982267 Chr.9: 6536154-6536154 on Build GRCh38
rs147762931 Chr.9: 6536208-6536208 on Build GRCh38
rs750518195 Chr.9: 6536088-6536088 on Build GRCh38
rs559923220 Chr.9: 6536084-6536084 on Build GRCh38
rs769606225 Chr.9: 6536218-6536218 on Build GRCh38
rs781057291 Chr.9: 6536128-6536128 on Build GRCh38
rs374969047 Chr.9: 6536071-6536071 on Build GRCh38
rs1054610342 Chr.9: 6536124-6536124 on Build GRCh38
rs764242927 Chr.9: 6536070-6536070 on Build GRCh38
rs552072250 Chr.9: 6536123-6536123 on Build GRCh38
rs367759326 Chr.9: 6536109-6536109 on Build GRCh38
rs377763882 Chr.9: 6536033-6536033 on Build GRCh38
rs369767254 Chr.9: 6536199-6536199 on Build GRCh38
rs771178748 Chr.9: 6536244-6536244 on Build GRCh38
rs767772993 Chr.9: 6536087-6536087 on Build GRCh38
rs367987650 Chr.9: 6536224-6536224 on Build GRCh38
rs780535258 Chr.9: 6536205-6536205 on Build GRCh38
rs200680938 Chr.9: 6536060-6536060 on Build GRCh38
rs754188922 Chr.9: 6536100-6536100 on Build GRCh38
rs775143096 Chr.9: 6536151-6536151 on Build GRCh38
rs190995960 Chr.9: 6536096-6536096 on Build GRCh38
rs868422652 Chr.9: 6536162-6536162 on Build GRCh38
rs752943013 Chr.9: 6536196-6536196 on Build GRCh38
rs386833568 Chr.9: 6536059-6536059 on Build GRCh38
rs760961076 Chr.9: 6536232-6536232 on Build GRCh38
rs370297328 Chr.9: 6536153-6536153 on Build GRCh38
rs756807420 Chr.9: 6536201-6536201 on Build GRCh38
rs1026865807 Chr.9: 6536248-6536248 on Build GRCh38
rs144939090 Chr.9: 6536229-6536229 on Build GRCh38
rs767018561 Chr.9: 6536167-6536167 on Build GRCh38
rs146939116 Chr.9: 6536228-6536228 on Build GRCh38
rs747637393 Chr.9: 6536035-6536035 on Build GRCh38
rs754593541 Chr.9: 6536027-6536027 on Build GRCh38
rs751089321 Chr.9: 6536200-6536200 on Build GRCh38
rs141152043 Chr.9: 6536219-6536219 on Build GRCh38
rs934940369 Chr.9: 6536122-6536122 on Build GRCh38

View Full Product Details