Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 158784551-158784816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EZR
Entrez Gene ID
7430
Gene Name
ezrin
Gene Aliases
CVIL, CVL, HEL-S-105, VIL2
UniGene
Hs.487027
Gene Chromosome Location
Chr.6: 158765741-158819424 on Build GRCh38
Transcripts
NM_001111077.1 NM_003379.4 XM_011536110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4440064 c.538C>T p.R180C Chr.6: 158784657-158784657 on Build GRCh38
7119814 c.549C>T p.L183L Chr.6: 158784646-158784646 on Build GRCh38
6455114 c.492C>T p.T164T Chr.6: 158784703-158784703 on Build GRCh38
6988831 c.513G>A p.R171R Chr.6: 158784682-158784682 on Build GRCh38
7230351 c.551+8C>T p.? Chr.6: 158784636-158784636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947215444 Chr.6: 158784748-158784748 on Build GRCh38
rs201151679 Chr.6: 158784584-158784584 on Build GRCh38
rs3103003 Chr.6: 158784636-158784636 on Build GRCh38
rs1044716896 Chr.6: 158784784-158784784 on Build GRCh38
rs780495451 Chr.6: 158784621-158784621 on Build GRCh38
rs755309400 Chr.6: 158784625-158784625 on Build GRCh38
rs762671735 Chr.6: 158784664-158784664 on Build GRCh38
rs752982231 Chr.6: 158784739-158784739 on Build GRCh38
rs780953809 Chr.6: 158784755-158784755 on Build GRCh38
rs745342152 Chr.6: 158784650-158784650 on Build GRCh38
rs9364995 Chr.6: 158784600-158784600 on Build GRCh38
rs766883419 Chr.6: 158784731-158784731 on Build GRCh38
rs768436026 Chr.6: 158784665-158784665 on Build GRCh38
rs749317363 Chr.6: 158784773-158784773 on Build GRCh38
rs370075029 Chr.6: 158784605-158784605 on Build GRCh38
rs3103004 Chr.6: 158784657-158784657 on Build GRCh38
rs757010720 Chr.6: 158784747-158784747 on Build GRCh38
rs770478466 Chr.6: 158784637-158784637 on Build GRCh38
rs1026474088 Chr.6: 158784594-158784594 on Build GRCh38
rs755435070 Chr.6: 158784734-158784734 on Build GRCh38
rs909035427 Chr.6: 158784780-158784780 on Build GRCh38
rs993874700 Chr.6: 158784586-158784586 on Build GRCh38
rs769373501 Chr.6: 158784769-158784769 on Build GRCh38
rs576184248 Chr.6: 158784771-158784771 on Build GRCh38
rs760694464 Chr.6: 158784722-158784722 on Build GRCh38
rs368572056 Chr.6: 158784738-158784738 on Build GRCh38
rs756652336 Chr.6: 158784617-158784617 on Build GRCh38
rs749635947 Chr.6: 158784623-158784623 on Build GRCh38
rs777295375 Chr.6: 158784629-158784629 on Build GRCh38
rs775467165 Chr.6: 158784595-158784595 on Build GRCh38
rs750531085 Chr.6: 158784710-158784710 on Build GRCh38
rs772403029 Chr.6: 158784742-158784743 on Build GRCh38
rs553782337 Chr.6: 158784792-158784792 on Build GRCh38
rs780898593 Chr.6: 158784648-158784648 on Build GRCh38
rs774015874 Chr.6: 158784683-158784683 on Build GRCh38
rs762139861 Chr.6: 158784694-158784694 on Build GRCh38
rs767690814 Chr.6: 158784704-158784704 on Build GRCh38
rs533508691 Chr.6: 158784599-158784599 on Build GRCh38
rs539532068 Chr.6: 158784656-158784656 on Build GRCh38
rs754302942 Chr.6: 158784732-158784732 on Build GRCh38
rs751752351 Chr.6: 158784604-158784604 on Build GRCh38
rs745397144 Chr.6: 158784759-158784759 on Build GRCh38
rs965087522 Chr.6: 158784628-158784628 on Build GRCh38
rs146339493 Chr.6: 158784785-158784785 on Build GRCh38

View Full Product Details