Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 17764850-17765119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEN1
Entrez Gene ID
348654
Gene Name
GEN1, Holliday junction 5' flap endonuclease
Gene Aliases
Gen
UniGene
Hs.467793
Gene Chromosome Location
Chr.2: 17753687-17785920 on Build GRCh38
Transcripts
NM_001130009.2 NM_182625.4 XM_005262613.4 XM_006712005.3 XM_011532820.2 XM_011532821.2 XM_011532822.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4001384 c.428A>G p.N143S Chr.2: 17764976-17764976 on Build GRCh38
4437029 c.426C>T p.L142L Chr.2: 17764974-17764974 on Build GRCh38
1168911 c.410_411insA p.M138fs*18 Chr.2: 17764958-17764959 on Build GRCh38
5620870 c.427A>G p.N143D Chr.2: 17764975-17764975 on Build GRCh38
6670470 c.394G>A p.A132T Chr.2: 17764942-17764942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750751476 Chr.2: 17764895-17764895 on Build GRCh38
rs752416254 Chr.2: 17764899-17764899 on Build GRCh38
rs777865001 Chr.2: 17765019-17765019 on Build GRCh38
rs760365006 Chr.2: 17764900-17764900 on Build GRCh38
rs748447726 Chr.2: 17764933-17764933 on Build GRCh38
rs781252989 Chr.2: 17764930-17764930 on Build GRCh38
rs778832705 Chr.2: 17764923-17764923 on Build GRCh38
rs761577393 Chr.2: 17765069-17765069 on Build GRCh38
rs760439371 Chr.2: 17764990-17764990 on Build GRCh38
rs201825923 Chr.2: 17765043-17765043 on Build GRCh38
rs765113183 Chr.2: 17764995-17764995 on Build GRCh38
rs752849386 Chr.2: 17765009-17765009 on Build GRCh38
rs200681783 Chr.2: 17764938-17764938 on Build GRCh38
rs562841392 Chr.2: 17764974-17764974 on Build GRCh38
rs759120118 Chr.2: 17764884-17764884 on Build GRCh38
rs770049577 Chr.2: 17764934-17764934 on Build GRCh38
rs759066905 Chr.2: 17764964-17764964 on Build GRCh38
rs765060095 Chr.2: 17765071-17765071 on Build GRCh38
rs530995407 Chr.2: 17765007-17765007 on Build GRCh38
rs750413085 Chr.2: 17765000-17765000 on Build GRCh38
rs763985908 Chr.2: 17764911-17764911 on Build GRCh38
rs757717475 Chr.2: 17765026-17765026 on Build GRCh38
rs201084834 Chr.2: 17764912-17764912 on Build GRCh38
rs139099020 Chr.2: 17765017-17765017 on Build GRCh38
rs755076488 Chr.2: 17764928-17764928 on Build GRCh38
rs546131542 Chr.2: 17764958-17764958 on Build GRCh38
rs922905215 Chr.2: 17765030-17765030 on Build GRCh38
rs773943586 Chr.2: 17764878-17764878 on Build GRCh38
rs143444612 Chr.2: 17765037-17765037 on Build GRCh38
rs150961964 Chr.2: 17765042-17765042 on Build GRCh38
rs990009959 Chr.2: 17764882-17764882 on Build GRCh38
rs933591876 Chr.2: 17765062-17765062 on Build GRCh38
rs768285016 Chr.2: 17765047-17765047 on Build GRCh38
rs776290211 Chr.2: 17765053-17765053 on Build GRCh38
rs759874288 Chr.2: 17764944-17764945 on Build GRCh38
rs749628704 Chr.2: 17765022-17765022 on Build GRCh38
rs984593945 Chr.2: 17764906-17764906 on Build GRCh38
rs368564254 Chr.2: 17765038-17765038 on Build GRCh38
rs767237406 Chr.2: 17764894-17764894 on Build GRCh38
rs542006404 Chr.2: 17764992-17764992 on Build GRCh38
rs16981869 Chr.2: 17764976-17764976 on Build GRCh38
rs773133354 Chr.2: 17765074-17765074 on Build GRCh38
rs753568841 Chr.2: 17764993-17764993 on Build GRCh38
rs199545866 Chr.2: 17764951-17764951 on Build GRCh38
rs767707920 Chr.2: 17765008-17765008 on Build GRCh38
rs912221893 Chr.2: 17764953-17764953 on Build GRCh38
rs111388449 Chr.2: 17764956-17764956 on Build GRCh38
rs751601024 Chr.2: 17764926-17764926 on Build GRCh38
rs749685885 Chr.2: 17764939-17764939 on Build GRCh38

View Full Product Details