Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120409475-120409732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSTL1
Entrez Gene ID
11167
Gene Name
follistatin like 1
Gene Aliases
FRP, FSL1, MIR198, OCC-1, OCC1, tsc36
UniGene
Hs.269512
Gene Chromosome Location
Chr.3: 120394214-120451071 on Build GRCh38
Transcripts
NM_007085.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4685772 c.364C>T p.R122* Chr.3: 120409630-120409630 on Build GRCh38
6504128 c.416T>G p.F139C Chr.3: 120409578-120409578 on Build GRCh38
3585828 c.375C>T p.I125I Chr.3: 120409619-120409619 on Build GRCh38
3585827 c.379C>A p.Q127K Chr.3: 120409615-120409615 on Build GRCh38
4836627 c.450C>A p.D150E Chr.3: 120409544-120409544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367753868 Chr.3: 120409506-120409506 on Build GRCh38
rs768600165 Chr.3: 120409650-120409650 on Build GRCh38
rs778663235 Chr.3: 120409541-120409541 on Build GRCh38
rs372311784 Chr.3: 120409704-120409704 on Build GRCh38
rs773106507 Chr.3: 120409560-120409560 on Build GRCh38
rs956909529 Chr.3: 120409649-120409649 on Build GRCh38
rs138210412 Chr.3: 120409690-120409690 on Build GRCh38
rs941550143 Chr.3: 120409564-120409564 on Build GRCh38
rs775869850 Chr.3: 120409571-120409571 on Build GRCh38
rs376856380 Chr.3: 120409623-120409623 on Build GRCh38
rs749566379 Chr.3: 120409648-120409648 on Build GRCh38
rs187480460 Chr.3: 120409558-120409558 on Build GRCh38
rs775814717 Chr.3: 120409643-120409643 on Build GRCh38
rs746741369 Chr.3: 120409702-120409702 on Build GRCh38
rs753507075 Chr.3: 120409624-120409624 on Build GRCh38
rs750151327 Chr.3: 120409578-120409578 on Build GRCh38
rs145669977 Chr.3: 120409642-120409642 on Build GRCh38
rs369259187 Chr.3: 120409627-120409627 on Build GRCh38
rs138798165 Chr.3: 120409570-120409570 on Build GRCh38
rs1013004553 Chr.3: 120409618-120409618 on Build GRCh38
rs138073641 Chr.3: 120409653-120409653 on Build GRCh38
rs376662981 Chr.3: 120409711-120409711 on Build GRCh38
rs554890949 Chr.3: 120409520-120409520 on Build GRCh38
rs113090248 Chr.3: 120409509-120409509 on Build GRCh38
rs757861275 Chr.3: 120409699-120409699 on Build GRCh38
rs879360534 Chr.3: 120409517-120409517 on Build GRCh38
rs762224570 Chr.3: 120409661-120409661 on Build GRCh38
rs113155123 Chr.3: 120409635-120409635 on Build GRCh38
rs765158611 Chr.3: 120409698-120409699 on Build GRCh38
rs201141663 Chr.3: 120409670-120409670 on Build GRCh38
rs368972992 Chr.3: 120409689-120409689 on Build GRCh38
rs1002607178 Chr.3: 120409559-120409559 on Build GRCh38
rs767998804 Chr.3: 120409669-120409669 on Build GRCh38
rs764301109 Chr.3: 120409586-120409586 on Build GRCh38
rs758420275 Chr.3: 120409609-120409609 on Build GRCh38
rs373512756 Chr.3: 120409680-120409680 on Build GRCh38
rs748013373 Chr.3: 120409629-120409629 on Build GRCh38
rs554312023 Chr.3: 120409626-120409626 on Build GRCh38
rs767763556 Chr.3: 120409587-120409587 on Build GRCh38
rs200383351 Chr.3: 120409692-120409692 on Build GRCh38
rs372544118 Chr.3: 120409633-120409633 on Build GRCh38
rs374398872 Chr.3: 120409676-120409676 on Build GRCh38
rs371453080 Chr.3: 120409556-120409556 on Build GRCh38
rs746073276 Chr.3: 120409562-120409562 on Build GRCh38
rs750868742 Chr.3: 120409607-120409607 on Build GRCh38
rs375964683 Chr.3: 120409500-120409500 on Build GRCh38
rs756472773 Chr.3: 120409614-120409614 on Build GRCh38

View Full Product Details