Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 40021252-40021481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83F
Entrez Gene ID
113828
Gene Name
family with sequence similarity 83 member F
Gene Aliases
-
UniGene
Hs.197680
Gene Chromosome Location
Chr.22: 39994949-40030039 on Build GRCh38
Transcripts
NM_138435.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
230820 c.889G>A p.E297K Chr.22: 40021399-40021399 on Build GRCh38
5711932 c.823C>A p.L275I Chr.22: 40021333-40021333 on Build GRCh38
6660234 c.858G>A p.T286T Chr.22: 40021368-40021368 on Build GRCh38
1682276 c.832G>A p.G278R Chr.22: 40021342-40021342 on Build GRCh38
6095297 c.865C>T p.R289W Chr.22: 40021375-40021375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754754707 Chr.22: 40021294-40021294 on Build GRCh38
rs765988859 Chr.22: 40021281-40021281 on Build GRCh38
rs900047302 Chr.22: 40021448-40021448 on Build GRCh38
rs144193825 Chr.22: 40021358-40021358 on Build GRCh38
rs772881620 Chr.22: 40021391-40021391 on Build GRCh38
rs749347650 Chr.22: 40021346-40021346 on Build GRCh38
rs995562237 Chr.22: 40021286-40021286 on Build GRCh38
rs868173254 Chr.22: 40021400-40021400 on Build GRCh38
rs567195386 Chr.22: 40021331-40021331 on Build GRCh38
rs956296780 Chr.22: 40021366-40021366 on Build GRCh38
rs781204642 Chr.22: 40021299-40021299 on Build GRCh38
rs764615099 Chr.22: 40021273-40021273 on Build GRCh38
rs199839917 Chr.22: 40021350-40021350 on Build GRCh38
rs772752278 Chr.22: 40021386-40021386 on Build GRCh38
rs752511860 Chr.22: 40021304-40021304 on Build GRCh38
rs750115679 Chr.22: 40021274-40021274 on Build GRCh38
rs756238744 Chr.22: 40021306-40021306 on Build GRCh38
rs369991191 Chr.22: 40021414-40021414 on Build GRCh38
rs1036963988 Chr.22: 40021421-40021421 on Build GRCh38
rs115837035 Chr.22: 40021368-40021368 on Build GRCh38
rs931142174 Chr.22: 40021441-40021441 on Build GRCh38
rs762647909 Chr.22: 40021278-40021278 on Build GRCh38
rs746044722 Chr.22: 40021450-40021450 on Build GRCh38
rs761040921 Chr.22: 40021375-40021375 on Build GRCh38
rs780334805 Chr.22: 40021458-40021458 on Build GRCh38
rs756804541 Chr.22: 40021452-40021454 on Build GRCh38
rs867836303 Chr.22: 40021342-40021342 on Build GRCh38
rs889750609 Chr.22: 40021343-40021343 on Build GRCh38
rs746245995 Chr.22: 40021363-40021363 on Build GRCh38
rs1032559846 Chr.22: 40021459-40021459 on Build GRCh38
rs577866074 Chr.22: 40021456-40021456 on Build GRCh38
rs772255056 Chr.22: 40021367-40021367 on Build GRCh38
rs142198025 Chr.22: 40021395-40021395 on Build GRCh38
rs769188379 Chr.22: 40021376-40021376 on Build GRCh38
rs147405763 Chr.22: 40021419-40021419 on Build GRCh38
rs752404354 Chr.22: 40021382-40021383 on Build GRCh38
rs751156757 Chr.22: 40021396-40021396 on Build GRCh38
rs751064389 Chr.22: 40021388-40021389 on Build GRCh38
rs764925220 Chr.22: 40021380-40021381 on Build GRCh38
rs762627037 Chr.22: 40021384-40021385 on Build GRCh38
rs756046306 Chr.22: 40021425-40021425 on Build GRCh38
rs139637555 Chr.22: 40021431-40021431 on Build GRCh38
rs145150338 Chr.22: 40021442-40021442 on Build GRCh38
rs762446693 Chr.22: 40021387-40021387 on Build GRCh38
rs777762843 Chr.22: 40021335-40021335 on Build GRCh38
rs763726408 Chr.22: 40021386-40021387 on Build GRCh38
rs751422717 Chr.22: 40021287-40021287 on Build GRCh38

View Full Product Details