Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 36581209-36581450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEZ2
Entrez Gene ID
9637
Gene Name
fasciculation and elongation protein zeta 2
Gene Aliases
HUM3CL
UniGene
Hs.258563
Gene Chromosome Location
Chr.2: 36552239-36598190 on Build GRCh38
Transcripts
NM_001042548.1 NM_005102.2 XM_006712153.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4943058 c.519G>A p.M173I Chr.2: 36581405-36581405 on Build GRCh38
2943438 c.613A>T p.S205C Chr.2: 36581311-36581311 on Build GRCh38
4904296 c.584C>T p.S195F Chr.2: 36581340-36581340 on Build GRCh38
3426412 c.516G>A p.M172I Chr.2: 36581408-36581408 on Build GRCh38
477364 c.610T>C p.S204P Chr.2: 36581314-36581314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779471628 Chr.2: 36581423-36581423 on Build GRCh38
rs773976422 Chr.2: 36581372-36581373 on Build GRCh38
rs773216559 Chr.2: 36581285-36581288 on Build GRCh38
rs753506178 Chr.2: 36581329-36581329 on Build GRCh38
rs547540281 Chr.2: 36581360-36581360 on Build GRCh38
rs757922872 Chr.2: 36581416-36581416 on Build GRCh38
rs1037549633 Chr.2: 36581271-36581271 on Build GRCh38
rs1002326059 Chr.2: 36581293-36581293 on Build GRCh38
rs759301228 Chr.2: 36581373-36581374 on Build GRCh38
rs757862888 Chr.2: 36581355-36581355 on Build GRCh38
rs762917944 Chr.2: 36581317-36581319 on Build GRCh38
rs771105597 Chr.2: 36581376-36581376 on Build GRCh38
rs770561244 Chr.2: 36581265-36581265 on Build GRCh38
rs199658522 Chr.2: 36581304-36581304 on Build GRCh38
rs563689370 Chr.2: 36581280-36581280 on Build GRCh38
rs150074766 Chr.2: 36581243-36581243 on Build GRCh38
rs377638928 Chr.2: 36581347-36581347 on Build GRCh38
rs749005530 Chr.2: 36581247-36581247 on Build GRCh38
rs535802106 Chr.2: 36581267-36581267 on Build GRCh38
rs777033656 Chr.2: 36581285-36581285 on Build GRCh38
rs770632912 Chr.2: 36581369-36581370 on Build GRCh38
rs373271952 Chr.2: 36581321-36581321 on Build GRCh38
rs201398630 Chr.2: 36581323-36581323 on Build GRCh38
rs549846320 Chr.2: 36581311-36581311 on Build GRCh38
rs766424883 Chr.2: 36581310-36581310 on Build GRCh38
rs781716577 Chr.2: 36581359-36581359 on Build GRCh38
rs755921231 Chr.2: 36581327-36581327 on Build GRCh38
rs374480931 Chr.2: 36581370-36581370 on Build GRCh38
rs762182519 Chr.2: 36581303-36581303 on Build GRCh38
rs750160244 Chr.2: 36581404-36581404 on Build GRCh38
rs909368451 Chr.2: 36581399-36581399 on Build GRCh38
rs778729731 Chr.2: 36581346-36581346 on Build GRCh38
rs749679504 Chr.2: 36581371-36581371 on Build GRCh38
rs759552008 Chr.2: 36581377-36581377 on Build GRCh38
rs755864062 Chr.2: 36581241-36581241 on Build GRCh38
rs775490423 Chr.2: 36581381-36581381 on Build GRCh38
rs781202356 Chr.2: 36581305-36581305 on Build GRCh38
rs960721244 Chr.2: 36581299-36581299 on Build GRCh38
rs760565426 Chr.2: 36581382-36581382 on Build GRCh38
rs35442829 Chr.2: 36581306-36581306 on Build GRCh38
rs775834019 Chr.2: 36581394-36581394 on Build GRCh38
rs747492840 Chr.2: 36581276-36581276 on Build GRCh38
rs748741967 Chr.2: 36581297-36581297 on Build GRCh38
rs757018959 Chr.2: 36581344-36581344 on Build GRCh38
rs763926773 Chr.2: 36581393-36581393 on Build GRCh38

View Full Product Details