Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41387415-41387676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC5
Entrez Gene ID
56915
Gene Name
exosome component 5
Gene Aliases
RRP41B, RRP46, Rrp46p, hRrp46p, p12B
UniGene
Hs.283741
Gene Chromosome Location
Chr.19: 41386370-41397351 on Build GRCh38
Transcripts
NM_020158.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5855880 c.568C>G p.R190G Chr.19: 41387561-41387561 on Build GRCh38
4078529 c.569G>A p.R190Q Chr.19: 41387560-41387560 on Build GRCh38
5570837 c.538G>A p.V180I Chr.19: 41387591-41387591 on Build GRCh38
4581083 c.584C>T p.S195F Chr.19: 41387545-41387545 on Build GRCh38
1393963 c.568C>T p.R190W Chr.19: 41387561-41387561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777915450 Chr.19: 41387550-41387550 on Build GRCh38
rs767345854 Chr.19: 41387603-41387603 on Build GRCh38
rs777702385 Chr.19: 41387614-41387614 on Build GRCh38
rs111391582 Chr.19: 41387510-41387510 on Build GRCh38
rs757030717 Chr.19: 41387625-41387625 on Build GRCh38
rs763844533 Chr.19: 41387635-41387635 on Build GRCh38
rs1036153412 Chr.19: 41387496-41387496 on Build GRCh38
rs747702828 Chr.19: 41387532-41387532 on Build GRCh38
rs778548687 Chr.19: 41387626-41387626 on Build GRCh38
rs151051277 Chr.19: 41387594-41387594 on Build GRCh38
rs1006470252 Chr.19: 41387459-41387459 on Build GRCh38
rs981471615 Chr.19: 41387487-41387487 on Build GRCh38
rs201468947 Chr.19: 41387596-41387596 on Build GRCh38
rs370300283 Chr.19: 41387472-41387472 on Build GRCh38
rs377456809 Chr.19: 41387561-41387561 on Build GRCh38
rs143662824 Chr.19: 41387567-41387567 on Build GRCh38
rs1008536968 Chr.19: 41387606-41387606 on Build GRCh38
rs770640185 Chr.19: 41387560-41387560 on Build GRCh38
rs991564510 Chr.19: 41387509-41387509 on Build GRCh38
rs754496082 Chr.19: 41387524-41387524 on Build GRCh38
rs772326064 Chr.19: 41387568-41387568 on Build GRCh38
rs772272912 Chr.19: 41387632-41387632 on Build GRCh38
rs370678317 Chr.19: 41387574-41387574 on Build GRCh38
rs777017215 Chr.19: 41387577-41387577 on Build GRCh38
rs201768777 Chr.19: 41387615-41387615 on Build GRCh38
rs759029443 Chr.19: 41387516-41387516 on Build GRCh38
rs764621753 Chr.19: 41387478-41387478 on Build GRCh38
rs112673788 Chr.19: 41387460-41387460 on Build GRCh38
rs199966819 Chr.19: 41387571-41387571 on Build GRCh38
rs867037676 Chr.19: 41387535-41387535 on Build GRCh38
rs752241723 Chr.19: 41387607-41387607 on Build GRCh38
rs760278018 Chr.19: 41387471-41387474 on Build GRCh38
rs754221443 Chr.19: 41387503-41387503 on Build GRCh38
rs757620072 Chr.19: 41387504-41387504 on Build GRCh38
rs756117524 Chr.19: 41387538-41387538 on Build GRCh38
rs745713122 Chr.19: 41387628-41387628 on Build GRCh38
rs749058888 Chr.19: 41387553-41387553 on Build GRCh38
rs887973062 Chr.19: 41387445-41387445 on Build GRCh38
rs762168533 Chr.19: 41387590-41387590 on Build GRCh38
rs140989771 Chr.19: 41387597-41387597 on Build GRCh38
rs764802562 Chr.19: 41387570-41387570 on Build GRCh38
rs370313944 Chr.19: 41387609-41387609 on Build GRCh38
rs766287840 Chr.19: 41387637-41387654 on Build GRCh38

View Full Product Details