Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123777118-123777391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM91A1
Entrez Gene ID
157769
Gene Name
family with sequence similarity 91 member A1
Gene Aliases
-
UniGene
Hs.459174
Gene Chromosome Location
Chr.8: 123768439-123815452 on Build GRCh38
Transcripts
NM_001317917.1 NM_001317918.1 NM_144963.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1454679 c.315T>C p.I105I Chr.8: 123777270-123777270 on Build GRCh38
5414926 c.341C>T p.S114L Chr.8: 123777296-123777296 on Build GRCh38
6209263 c.339T>C p.D113D Chr.8: 123777294-123777294 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768952447 Chr.8: 123777225-123777225 on Build GRCh38
rs1045142833 Chr.8: 123777207-123777207 on Build GRCh38
rs752274214 Chr.8: 123777340-123777340 on Build GRCh38
rs756895244 Chr.8: 123777209-123777209 on Build GRCh38
rs766540869 Chr.8: 123777247-123777247 on Build GRCh38
rs1019616931 Chr.8: 123777319-123777319 on Build GRCh38
rs948465935 Chr.8: 123777187-123777187 on Build GRCh38
rs7012004 Chr.8: 123777177-123777177 on Build GRCh38
rs779124785 Chr.8: 123777368-123777369 on Build GRCh38
rs748361309 Chr.8: 123777246-123777246 on Build GRCh38
rs776937356 Chr.8: 123777333-123777333 on Build GRCh38
rs751181140 Chr.8: 123777353-123777353 on Build GRCh38
rs1049053855 Chr.8: 123777281-123777281 on Build GRCh38
rs934592180 Chr.8: 123777171-123777171 on Build GRCh38
rs749127236 Chr.8: 123777224-123777224 on Build GRCh38
rs73335687 Chr.8: 123777143-123777143 on Build GRCh38
rs996916104 Chr.8: 123777349-123777349 on Build GRCh38
rs766797715 Chr.8: 123777331-123777331 on Build GRCh38
rs112538103 Chr.8: 123777260-123777260 on Build GRCh38
rs930163523 Chr.8: 123777172-123777172 on Build GRCh38
rs754002969 Chr.8: 123777329-123777333 on Build GRCh38
rs1034977361 Chr.8: 123777279-123777279 on Build GRCh38
rs976989929 Chr.8: 123777141-123777141 on Build GRCh38
rs142455870 Chr.8: 123777300-123777300 on Build GRCh38
rs879669002 Chr.8: 123777203-123777203 on Build GRCh38
rs1044165721 Chr.8: 123777193-123777193 on Build GRCh38
rs773226451 Chr.8: 123777285-123777285 on Build GRCh38
rs771027916 Chr.8: 123777342-123777342 on Build GRCh38
rs765636964 Chr.8: 123777339-123777339 on Build GRCh38
rs761181662 Chr.8: 123777294-123777294 on Build GRCh38
rs77170970 Chr.8: 123777180-123777180 on Build GRCh38
rs755127702 Chr.8: 123777355-123777355 on Build GRCh38
rs1003873876 Chr.8: 123777232-123777232 on Build GRCh38
rs190754516 Chr.8: 123777337-123777337 on Build GRCh38
rs761439374 Chr.8: 123777359-123777359 on Build GRCh38
rs578022402 Chr.8: 123777211-123777211 on Build GRCh38
rs772192079 Chr.8: 123777273-123777273 on Build GRCh38
rs767039912 Chr.8: 123777361-123777361 on Build GRCh38
rs774695810 Chr.8: 123777237-123777237 on Build GRCh38

View Full Product Details