Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19132190-19132449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR14
Entrez Gene ID
8220
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
DGCR13, DGS-H, DGS-I, DGSH, DGSI, ES2, Es2el
UniGene
Hs.517407
Gene Chromosome Location
Chr.22: 19130279-19144726 on Build GRCh38
Transcripts
NM_022719.2 NR_134304.1 NM_053006.4

Gene Symbol
TSSK2
Entrez Gene ID
23617
Gene Name
testis specific serine kinase 2
Gene Aliases
DGS-G, SPOGA2, STK22B, TSK2
UniGene
Hs.694070
Gene Chromosome Location
Chr.22: 19130808-19132623 on Build GRCh38
Transcripts
NM_022719.2 NR_134304.1 NM_053006.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6810767 c.965G>A p.R322Q Chr.22: 19132364-19132364 on Build GRCh38
7395529 c.987C>A p.N329K Chr.22: 19132386-19132386 on Build GRCh38
4561075 c.868G>A p.E290K Chr.22: 19132267-19132267 on Build GRCh38
1414854 c.1047C>T p.S349S Chr.22: 19132446-19132446 on Build GRCh38
5858893 c.911G>A p.G304D Chr.22: 19132310-19132310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759143051 Chr.22: 19132281-19132281 on Build GRCh38
rs762012303 Chr.22: 19132292-19132292 on Build GRCh38
rs764514103 Chr.22: 19132218-19132219 on Build GRCh38
rs139396065 Chr.22: 19132317-19132317 on Build GRCh38
rs762070638 Chr.22: 19132316-19132327 on Build GRCh38
rs767669673 Chr.22: 19132328-19132339 on Build GRCh38
rs757352938 Chr.22: 19132237-19132237 on Build GRCh38
rs753168306 Chr.22: 19132213-19132213 on Build GRCh38
rs781106529 Chr.22: 19132421-19132421 on Build GRCh38
rs776300390 Chr.22: 19132280-19132280 on Build GRCh38
rs141616852 Chr.22: 19132265-19132265 on Build GRCh38
rs1052763 Chr.22: 19132238-19132238 on Build GRCh38
rs1033214507 Chr.22: 19132392-19132392 on Build GRCh38
rs751715581 Chr.22: 19132230-19132230 on Build GRCh38
rs758996674 Chr.22: 19132219-19132219 on Build GRCh38
rs777773500 Chr.22: 19132327-19132327 on Build GRCh38
rs146810069 Chr.22: 19132380-19132380 on Build GRCh38
rs774634166 Chr.22: 19132236-19132237 on Build GRCh38
rs752596217 Chr.22: 19132321-19132321 on Build GRCh38
rs750597559 Chr.22: 19132389-19132389 on Build GRCh38
rs140549220 Chr.22: 19132414-19132414 on Build GRCh38
rs746537468 Chr.22: 19132271-19132271 on Build GRCh38
rs374703905 Chr.22: 19132266-19132266 on Build GRCh38
rs757230340 Chr.22: 19132416-19132416 on Build GRCh38
rs150913850 Chr.22: 19132288-19132288 on Build GRCh38
rs200322808 Chr.22: 19132251-19132251 on Build GRCh38
rs373650594 Chr.22: 19132328-19132328 on Build GRCh38
rs745355051 Chr.22: 19132363-19132363 on Build GRCh38
rs374835818 Chr.22: 19132287-19132287 on Build GRCh38
rs754841350 Chr.22: 19132248-19132248 on Build GRCh38
rs758245828 Chr.22: 19132407-19132407 on Build GRCh38
rs746046655 Chr.22: 19132239-19132239 on Build GRCh38
rs766637203 Chr.22: 19132397-19132397 on Build GRCh38
rs760659207 Chr.22: 19132393-19132393 on Build GRCh38
rs751941684 Chr.22: 19132290-19132290 on Build GRCh38
rs753942362 Chr.22: 19132398-19132398 on Build GRCh38
rs768106552 Chr.22: 19132387-19132387 on Build GRCh38
rs775019033 Chr.22: 19132381-19132381 on Build GRCh38
rs185310637 Chr.22: 19132312-19132312 on Build GRCh38
rs139504972 Chr.22: 19132263-19132263 on Build GRCh38
rs989638459 Chr.22: 19132346-19132346 on Build GRCh38
rs1052773 Chr.22: 19132425-19132425 on Build GRCh38
rs769560398 Chr.22: 19132364-19132364 on Build GRCh38
rs777385407 Chr.22: 19132413-19132413 on Build GRCh38
rs150037057 Chr.22: 19132320-19132320 on Build GRCh38
rs764779842 Chr.22: 19132282-19132282 on Build GRCh38
rs17853776 Chr.22: 19132310-19132310 on Build GRCh38
rs551983510 Chr.22: 19132227-19132227 on Build GRCh38
rs376847235 Chr.22: 19132386-19132386 on Build GRCh38
rs549857607 Chr.22: 19132311-19132311 on Build GRCh38
rs140125132 Chr.22: 19132333-19132333 on Build GRCh38
rs770453644 Chr.22: 19132332-19132332 on Build GRCh38
rs143749090 Chr.22: 19132371-19132371 on Build GRCh38
rs371353822 Chr.22: 19132279-19132279 on Build GRCh38
rs748715816 Chr.22: 19132368-19132368 on Build GRCh38
rs756444543 Chr.22: 19132313-19132313 on Build GRCh38
rs145304535 Chr.22: 19132323-19132323 on Build GRCh38

View Full Product Details